Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66494373-66494608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAPC5
Entrez Gene ID
10302
Gene Name
small nuclear RNA activating complex polypeptide 5
Gene Aliases
SNAP19
UniGene
Hs.30174
Gene Chromosome Location
Chr.15: 66490328-66497808 on Build GRCh38
Transcripts
NM_006049.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5170950 c.270_272delGGA p.E95delE Chr.15: 66494461-66494463 on Build GRCh38
212348 c.254C>G p.T85R Chr.15: 66494479-66494479 on Build GRCh38
6231501 c.247C>T p.H83Y Chr.15: 66494486-66494486 on Build GRCh38
6785966 c.283G>T p.E95* Chr.15: 66494450-66494450 on Build GRCh38
3706697 c.199A>G p.N67D Chr.15: 66494534-66494534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144538572 Chr.15: 66494479-66494479 on Build GRCh38
rs756614440 Chr.15: 66494414-66494414 on Build GRCh38
rs549029337 Chr.15: 66494411-66494411 on Build GRCh38
rs998947972 Chr.15: 66494427-66494427 on Build GRCh38
rs772409883 Chr.15: 66494532-66494532 on Build GRCh38
rs758455721 Chr.15: 66494461-66494461 on Build GRCh38
rs1019344790 Chr.15: 66494437-66494437 on Build GRCh38
rs776097640 Chr.15: 66494549-66494549 on Build GRCh38
rs1030584678 Chr.15: 66494456-66494456 on Build GRCh38
rs559628527 Chr.15: 66494412-66494412 on Build GRCh38
rs996658026 Chr.15: 66494457-66494457 on Build GRCh38
rs754124299 Chr.15: 66494429-66494429 on Build GRCh38
rs764183738 Chr.15: 66494415-66494415 on Build GRCh38
rs767892732 Chr.15: 66494406-66494406 on Build GRCh38
rs776635295 Chr.15: 66494461-66494463 on Build GRCh38
rs774677741 Chr.15: 66494485-66494485 on Build GRCh38
rs760105986 Chr.15: 66494399-66494399 on Build GRCh38
rs150019625 Chr.15: 66494444-66494444 on Build GRCh38
rs779200180 Chr.15: 66494450-66494450 on Build GRCh38
rs754107883 Chr.15: 66494578-66494578 on Build GRCh38
rs768499654 Chr.15: 66494478-66494478 on Build GRCh38
rs764575654 Chr.15: 66494562-66494562 on Build GRCh38
rs551596192 Chr.15: 66494504-66494504 on Build GRCh38
rs368120374 Chr.15: 66494542-66494542 on Build GRCh38
rs908497016 Chr.15: 66494521-66494521 on Build GRCh38
rs1028164354 Chr.15: 66494466-66494466 on Build GRCh38
rs746937861 Chr.15: 66494409-66494412 on Build GRCh38
rs139309951 Chr.15: 66494443-66494443 on Build GRCh38
rs771068116 Chr.15: 66494461-66494466 on Build GRCh38
rs531032132 Chr.15: 66494533-66494533 on Build GRCh38
rs373687057 Chr.15: 66494460-66494460 on Build GRCh38
rs913012792 Chr.15: 66494589-66494589 on Build GRCh38
rs747859044 Chr.15: 66494481-66494481 on Build GRCh38
rs749272921 Chr.15: 66494460-66494461 on Build GRCh38
rs760889276 Chr.15: 66494559-66494559 on Build GRCh38
rs768544844 Chr.15: 66494444-66494445 on Build GRCh38
rs796052169 Chr.15: 66494510-66494510 on Build GRCh38
rs759880147 Chr.15: 66494565-66494566 on Build GRCh38
rs1018328657 Chr.15: 66494534-66494534 on Build GRCh38
rs763732658 Chr.15: 66494405-66494406 on Build GRCh38
rs780058474 Chr.15: 66494471-66494471 on Build GRCh38
rs571352833 Chr.15: 66494528-66494528 on Build GRCh38

View Full Product Details