Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3289617-3289864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF263
Entrez Gene ID
10127
Gene Name
zinc finger protein 263
Gene Aliases
FPM315, ZKSCAN12, ZSCAN44
UniGene
Hs.611475
Gene Chromosome Location
Chr.16: 3283183-3301401 on Build GRCh38
Transcripts
NM_005741.4 XM_006720831.1 XM_006720832.1 XM_006720833.1 XM_006720834.3 XM_011522344.1 XM_017022850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3817933 c.1193G>T p.R398I Chr.16: 3289699-3289699 on Build GRCh38
2919104 c.1153delA p.N386fs*8 Chr.16: 3289659-3289659 on Build GRCh38
3969558 c.1259G>A p.R420H Chr.16: 3289765-3289765 on Build GRCh38
3402312 c.1188C>A p.H396Q Chr.16: 3289694-3289694 on Build GRCh38
1233779 c.1211G>T p.R404I Chr.16: 3289717-3289717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917438873 Chr.16: 3289743-3289743 on Build GRCh38
rs763241212 Chr.16: 3289702-3289702 on Build GRCh38
rs371017559 Chr.16: 3289764-3289764 on Build GRCh38
rs746994545 Chr.16: 3289648-3289648 on Build GRCh38
rs779791895 Chr.16: 3289638-3289638 on Build GRCh38
rs771274938 Chr.16: 3289803-3289803 on Build GRCh38
rs1050442564 Chr.16: 3289800-3289800 on Build GRCh38
rs760155666 Chr.16: 3289714-3289714 on Build GRCh38
rs561763622 Chr.16: 3289712-3289712 on Build GRCh38
rs759833600 Chr.16: 3289819-3289819 on Build GRCh38
rs748082036 Chr.16: 3289776-3289776 on Build GRCh38
rs143673234 Chr.16: 3289847-3289847 on Build GRCh38
rs756739328 Chr.16: 3289741-3289741 on Build GRCh38
rs958430077 Chr.16: 3289660-3289660 on Build GRCh38
rs746793559 Chr.16: 3289647-3289647 on Build GRCh38
rs765822647 Chr.16: 3289790-3289790 on Build GRCh38
rs767872209 Chr.16: 3289826-3289826 on Build GRCh38
rs766902363 Chr.16: 3289706-3289706 on Build GRCh38
rs748177334 Chr.16: 3289676-3289676 on Build GRCh38
rs773408212 Chr.16: 3289782-3289782 on Build GRCh38
rs944775366 Chr.16: 3289840-3289840 on Build GRCh38
rs750116677 Chr.16: 3289751-3289751 on Build GRCh38
rs773602336 Chr.16: 3289694-3289694 on Build GRCh38
rs767915747 Chr.16: 3289721-3289721 on Build GRCh38
rs374333605 Chr.16: 3289792-3289792 on Build GRCh38
rs910666389 Chr.16: 3289836-3289836 on Build GRCh38
rs189669184 Chr.16: 3289738-3289738 on Build GRCh38
rs776835539 Chr.16: 3289663-3289663 on Build GRCh38
rs200998380 Chr.16: 3289726-3289726 on Build GRCh38
rs993018733 Chr.16: 3289736-3289736 on Build GRCh38
rs768461124 Chr.16: 3289651-3289651 on Build GRCh38
rs758023508 Chr.16: 3289752-3289752 on Build GRCh38
rs764684677 Chr.16: 3289746-3289746 on Build GRCh38
rs774649019 Chr.16: 3289809-3289809 on Build GRCh38
rs902551253 Chr.16: 3289808-3289808 on Build GRCh38
rs754733663 Chr.16: 3289769-3289769 on Build GRCh38
rs769872173 Chr.16: 3289688-3289688 on Build GRCh38
rs149007034 Chr.16: 3289765-3289765 on Build GRCh38
rs770545096 Chr.16: 3289711-3289711 on Build GRCh38
rs769807273 Chr.16: 3289780-3289780 on Build GRCh38
rs781003073 Chr.16: 3289773-3289773 on Build GRCh38

View Full Product Details