Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 61480077-61480344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX4
Entrez Gene ID
9496
Gene Name
T-box 4
Gene Aliases
ICPPS, SPS
UniGene
Hs.143907
Gene Chromosome Location
Chr.17: 61452418-61485110 on Build GRCh38
Transcripts
NM_001321120.1 NM_018488.3 XM_011525490.2 XM_011525491.2 XM_011525495.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5730928 c.819C>T p.S273S Chr.17: 61480117-61480117 on Build GRCh38
7333438 c.790-6C>G p.? Chr.17: 61480082-61480082 on Build GRCh38
285614 c.1021+7C>T p.? Chr.17: 61480326-61480326 on Build GRCh38
122013 c.942G>A p.A314A Chr.17: 61480240-61480240 on Build GRCh38
1290738 c.839T>A p.I280N Chr.17: 61480137-61480137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201565504 Chr.17: 61480240-61480240 on Build GRCh38
rs150233337 Chr.17: 61480283-61480283 on Build GRCh38
rs142327430 Chr.17: 61480174-61480174 on Build GRCh38
rs761870318 Chr.17: 61480173-61480173 on Build GRCh38
rs370805904 Chr.17: 61480276-61480276 on Build GRCh38
rs751264961 Chr.17: 61480201-61480201 on Build GRCh38
rs759150888 Chr.17: 61480161-61480161 on Build GRCh38
rs950842943 Chr.17: 61480257-61480257 on Build GRCh38
rs750033041 Chr.17: 61480135-61480135 on Build GRCh38
rs200387033 Chr.17: 61480245-61480245 on Build GRCh38
rs550334195 Chr.17: 61480153-61480153 on Build GRCh38
rs772552261 Chr.17: 61480255-61480255 on Build GRCh38
rs73334576 Chr.17: 61480142-61480142 on Build GRCh38
rs551973242 Chr.17: 61480244-61480244 on Build GRCh38
rs766300906 Chr.17: 61480139-61480140 on Build GRCh38
rs372765864 Chr.17: 61480117-61480117 on Build GRCh38
rs767809364 Chr.17: 61480285-61480285 on Build GRCh38
rs371242130 Chr.17: 61480238-61480238 on Build GRCh38
rs751211966 Chr.17: 61480271-61480271 on Build GRCh38
rs777644345 Chr.17: 61480237-61480237 on Build GRCh38
rs145889644 Chr.17: 61480102-61480102 on Build GRCh38
rs199780097 Chr.17: 61480261-61480261 on Build GRCh38
rs749954407 Chr.17: 61480180-61480180 on Build GRCh38
rs202061937 Chr.17: 61480230-61480230 on Build GRCh38
rs765917350 Chr.17: 61480268-61480268 on Build GRCh38
rs760566882 Chr.17: 61480166-61480166 on Build GRCh38
rs113899943 Chr.17: 61480225-61480225 on Build GRCh38
rs778553600 Chr.17: 61480126-61480126 on Build GRCh38
rs778365568 Chr.17: 61480317-61480317 on Build GRCh38
rs193204039 Chr.17: 61480160-61480160 on Build GRCh38
rs769106353 Chr.17: 61480144-61480144 on Build GRCh38
rs3744438 Chr.17: 61480239-61480239 on Build GRCh38
rs143895322 Chr.17: 61480181-61480181 on Build GRCh38
rs775385092 Chr.17: 61480165-61480165 on Build GRCh38
rs572207593 Chr.17: 61480103-61480103 on Build GRCh38
rs541224543 Chr.17: 61480121-61480121 on Build GRCh38
rs564100262 Chr.17: 61480138-61480138 on Build GRCh38
rs765251401 Chr.17: 61480246-61480246 on Build GRCh38
rs942011345 Chr.17: 61480315-61480315 on Build GRCh38
rs773396940 Chr.17: 61480154-61480154 on Build GRCh38
rs148616062 Chr.17: 61480213-61480213 on Build GRCh38
rs746131244 Chr.17: 61480241-61480241 on Build GRCh38
rs766046321 Chr.17: 61480194-61480194 on Build GRCh38
rs917982090 Chr.17: 61480107-61480107 on Build GRCh38
rs756535438 Chr.17: 61480304-61480304 on Build GRCh38
rs201395852 Chr.17: 61480155-61480155 on Build GRCh38
rs141188668 Chr.17: 61480219-61480219 on Build GRCh38
rs747409546 Chr.17: 61480143-61480143 on Build GRCh38
rs577197197 Chr.17: 61480303-61480303 on Build GRCh38
rs748587793 Chr.17: 61480220-61480220 on Build GRCh38
rs764158066 Chr.17: 61480116-61480116 on Build GRCh38

View Full Product Details