Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 52088271-52088510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP64
Entrez Gene ID
55734
Gene Name
ZFP64 zinc finger protein
Gene Aliases
ZNF338
UniGene
Hs.473082
Gene Chromosome Location
Chr.20: 52084011-52191985 on Build GRCh38
Transcripts
NM_001319146.1 NM_199427.2 XM_006723822.1 XM_017027945.1 XM_017027947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2763162 c.1155G>A p.P385P Chr.20: 52088465-52088465 on Build GRCh38
4099468 c.1132C>T p.R378W Chr.20: 52088488-52088488 on Build GRCh38
2763161 c.1207G>A p.V403I Chr.20: 52088413-52088413 on Build GRCh38
4742644 c.1187G>A p.R396H Chr.20: 52088433-52088433 on Build GRCh38
6585467 c.1122G>A p.K374K Chr.20: 52088498-52088498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003446473 Chr.20: 52088434-52088434 on Build GRCh38
rs1054467505 Chr.20: 52088371-52088371 on Build GRCh38
rs6096740 Chr.20: 52088417-52088417 on Build GRCh38
rs566185172 Chr.20: 52088377-52088377 on Build GRCh38
rs370373546 Chr.20: 52088466-52088466 on Build GRCh38
rs769133483 Chr.20: 52088355-52088355 on Build GRCh38
rs763965506 Chr.20: 52088404-52088404 on Build GRCh38
rs746997556 Chr.20: 52088433-52088433 on Build GRCh38
rs78754336 Chr.20: 52088393-52088393 on Build GRCh38
rs773444837 Chr.20: 52088370-52088370 on Build GRCh38
rs776706383 Chr.20: 52088358-52088358 on Build GRCh38
rs377689307 Chr.20: 52088444-52088444 on Build GRCh38
rs140282228 Chr.20: 52088414-52088414 on Build GRCh38
rs765364240 Chr.20: 52088368-52088368 on Build GRCh38
rs780538137 Chr.20: 52088423-52088423 on Build GRCh38
rs760668340 Chr.20: 52088488-52088488 on Build GRCh38
rs773608495 Chr.20: 52088465-52088465 on Build GRCh38
rs958622690 Chr.20: 52088303-52088303 on Build GRCh38
rs756936017 Chr.20: 52088410-52088410 on Build GRCh38
rs755846396 Chr.20: 52088403-52088403 on Build GRCh38
rs777081948 Chr.20: 52088443-52088443 on Build GRCh38
rs770113990 Chr.20: 52088453-52088453 on Build GRCh38
rs767529907 Chr.20: 52088384-52088384 on Build GRCh38
rs79095092 Chr.20: 52088462-52088462 on Build GRCh38
rs1000573900 Chr.20: 52088298-52088298 on Build GRCh38
rs753366493 Chr.20: 52088408-52088408 on Build GRCh38
rs899728492 Chr.20: 52088424-52088424 on Build GRCh38
rs373948550 Chr.20: 52088413-52088413 on Build GRCh38
rs768833411 Chr.20: 52088441-52088441 on Build GRCh38
rs762096768 Chr.20: 52088365-52088365 on Build GRCh38
rs775231013 Chr.20: 52088487-52088487 on Build GRCh38
rs747308404 Chr.20: 52088350-52088350 on Build GRCh38

View Full Product Details