Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131962472-131962737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL6
Entrez Gene ID
23305
Gene Name
acyl-CoA synthetase long-chain family member 6
Gene Aliases
ACS2, FACL6, LACS 6, LACS2, LACS5
UniGene
Hs.14945
Gene Chromosome Location
Chr.5: 131949973-132012068 on Build GRCh38
Transcripts
NM_001009185.2 NM_001205247.1 NM_001205248.1 NM_001205250.1 NM_001205251.1 NM_015256.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3941022 c.1819C>A p.P607T Chr.5: 131962573-131962573 on Build GRCh38
5008261 c.1796C>T p.A599V Chr.5: 131962566-131962566 on Build GRCh38
3849993 c.1752G>A p.A584A Chr.5: 131962565-131962565 on Build GRCh38
3849992 c.1827G>A p.A609A Chr.5: 131962565-131962565 on Build GRCh38
4656987 c.1754C>A p.P585H Chr.5: 131962608-131962608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761892148 Chr.5: 131962505-131962505 on Build GRCh38
rs1025026634 Chr.5: 131962648-131962648 on Build GRCh38
rs763597231 Chr.5: 131962634-131962634 on Build GRCh38
rs370745130 Chr.5: 131962607-131962607 on Build GRCh38
rs768946592 Chr.5: 131962626-131962626 on Build GRCh38
rs199688979 Chr.5: 131962652-131962652 on Build GRCh38
rs759417478 Chr.5: 131962710-131962710 on Build GRCh38
rs749724281 Chr.5: 131962610-131962610 on Build GRCh38
rs371575337 Chr.5: 131962510-131962510 on Build GRCh38
rs767581273 Chr.5: 131962509-131962509 on Build GRCh38
rs148922137 Chr.5: 131962577-131962577 on Build GRCh38
rs540883722 Chr.5: 131962599-131962599 on Build GRCh38
rs932721886 Chr.5: 131962547-131962547 on Build GRCh38
rs199637377 Chr.5: 131962578-131962578 on Build GRCh38
rs906232409 Chr.5: 131962654-131962654 on Build GRCh38
rs766688213 Chr.5: 131962705-131962705 on Build GRCh38
rs754815249 Chr.5: 131962573-131962573 on Build GRCh38
rs148539410 Chr.5: 131962580-131962580 on Build GRCh38
rs147880318 Chr.5: 131962566-131962566 on Build GRCh38
rs765076023 Chr.5: 131962715-131962715 on Build GRCh38
rs773386832 Chr.5: 131962678-131962678 on Build GRCh38
rs144652904 Chr.5: 131962527-131962527 on Build GRCh38
rs760657135 Chr.5: 131962698-131962698 on Build GRCh38
rs934884336 Chr.5: 131962682-131962682 on Build GRCh38
rs183511238 Chr.5: 131962582-131962582 on Build GRCh38
rs766487192 Chr.5: 131962565-131962565 on Build GRCh38
rs748644268 Chr.5: 131962646-131962646 on Build GRCh38
rs776926152 Chr.5: 131962707-131962707 on Build GRCh38
rs922637486 Chr.5: 131962529-131962529 on Build GRCh38
rs758826280 Chr.5: 131962576-131962576 on Build GRCh38
rs945668535 Chr.5: 131962512-131962512 on Build GRCh38

View Full Product Details