Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43226150-43226411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP36L2
Entrez Gene ID
678
Gene Name
ZFP36 ring finger protein like 2
Gene Aliases
BRF2, ERF-2, ERF2, RNF162C, TIS11D
UniGene
Hs.503093
Gene Chromosome Location
Chr.2: 43222402-43226606 on Build GRCh38
Transcripts
NM_006887.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
442913 c.5C>G p.S2W Chr.2: 43226311-43226311 on Build GRCh38
5431123 c.42C>A p.F14L Chr.2: 43226274-43226274 on Build GRCh38
5973800 c.47G>A p.C16Y Chr.2: 43226269-43226269 on Build GRCh38
4571483 c.46T>G p.C16G Chr.2: 43226270-43226270 on Build GRCh38
6534437 c.34G>C p.V12L Chr.2: 43226282-43226282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004447416 Chr.2: 43226390-43226390 on Build GRCh38
rs986980625 Chr.2: 43226392-43226392 on Build GRCh38
rs748071714 Chr.2: 43226236-43226236 on Build GRCh38
rs527528093 Chr.2: 43226329-43226329 on Build GRCh38
rs776940187 Chr.2: 43226269-43226269 on Build GRCh38
rs753420770 Chr.2: 43226300-43226300 on Build GRCh38
rs923696405 Chr.2: 43226171-43226171 on Build GRCh38
rs377079504 Chr.2: 43226366-43226366 on Build GRCh38
rs374962654 Chr.2: 43226327-43226327 on Build GRCh38
rs975788560 Chr.2: 43226170-43226170 on Build GRCh38
rs368801337 Chr.2: 43226328-43226328 on Build GRCh38
rs765835005 Chr.2: 43226367-43226367 on Build GRCh38
rs1053283775 Chr.2: 43226176-43226176 on Build GRCh38
rs934790913 Chr.2: 43226173-43226173 on Build GRCh38
rs201662483 Chr.2: 43226316-43226316 on Build GRCh38
rs759845224 Chr.2: 43226271-43226271 on Build GRCh38
rs754616453 Chr.2: 43226303-43226303 on Build GRCh38
rs746602454 Chr.2: 43226183-43226183 on Build GRCh38
rs1038456482 Chr.2: 43226357-43226357 on Build GRCh38
rs372899666 Chr.2: 43226215-43226215 on Build GRCh38
rs754618961 Chr.2: 43226239-43226239 on Build GRCh38
rs765886873 Chr.2: 43226294-43226294 on Build GRCh38
rs1039333646 Chr.2: 43226218-43226218 on Build GRCh38
rs777898706 Chr.2: 43226259-43226259 on Build GRCh38
rs754742107 Chr.2: 43226233-43226233 on Build GRCh38
rs990682911 Chr.2: 43226346-43226346 on Build GRCh38
rs371516427 Chr.2: 43226287-43226287 on Build GRCh38
rs771197019 Chr.2: 43226268-43226268 on Build GRCh38
rs531595158 Chr.2: 43226292-43226292 on Build GRCh38
rs979075751 Chr.2: 43226374-43226374 on Build GRCh38
rs781456423 Chr.2: 43226334-43226334 on Build GRCh38
rs774782250 Chr.2: 43226363-43226363 on Build GRCh38
rs935095450 Chr.2: 43226385-43226385 on Build GRCh38
rs771072304 Chr.2: 43226331-43226331 on Build GRCh38
rs746050063 Chr.2: 43226338-43226338 on Build GRCh38
rs752405606 Chr.2: 43226323-43226323 on Build GRCh38
rs201000261 Chr.2: 43226360-43226360 on Build GRCh38
rs563686680 Chr.2: 43226261-43226261 on Build GRCh38
rs923636776 Chr.2: 43226376-43226376 on Build GRCh38
rs368634680 Chr.2: 43226207-43226207 on Build GRCh38
rs888706802 Chr.2: 43226384-43226384 on Build GRCh38
rs112545275 Chr.2: 43226263-43226263 on Build GRCh38
rs574206300 Chr.2: 43226327-43226341 on Build GRCh38
rs775817612 Chr.2: 43226350-43226350 on Build GRCh38
rs775736445 Chr.2: 43226282-43226282 on Build GRCh38
rs769122759 Chr.2: 43226362-43226362 on Build GRCh38
rs770158304 Chr.2: 43226273-43226273 on Build GRCh38
rs994856050 Chr.2: 43226240-43226240 on Build GRCh38
rs771884716 Chr.2: 43226256-43226256 on Build GRCh38
rs111586080 Chr.2: 43226264-43226264 on Build GRCh38
rs542193287 Chr.2: 43226304-43226304 on Build GRCh38
rs754597699 Chr.2: 43226198-43226198 on Build GRCh38
rs763421001 Chr.2: 43226285-43226285 on Build GRCh38
rs902996805 Chr.2: 43226295-43226295 on Build GRCh38
rs779676950 Chr.2: 43226221-43226221 on Build GRCh38
rs375492308 Chr.2: 43226231-43226231 on Build GRCh38
rs372600331 Chr.2: 43226339-43226339 on Build GRCh38
rs764473720 Chr.2: 43226286-43226286 on Build GRCh38
rs182373932 Chr.2: 43226235-43226235 on Build GRCh38
rs890660771 Chr.2: 43226184-43226184 on Build GRCh38
rs967649038 Chr.2: 43226370-43226370 on Build GRCh38

View Full Product Details