Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45345794-45346067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TESK2
Entrez Gene ID
10420
Gene Name
testis-specific kinase 2
Gene Aliases
-
UniGene
Hs.591499
Gene Chromosome Location
Chr.1: 45343883-45491342 on Build GRCh38
Transcripts
NM_001320800.1 NM_007170.2 XM_005270355.3 XM_006710287.2 XM_011540497.1 XM_011540500.2 XM_017000047.1 XM_017000048.1 XM_017000049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4781292 c.889A>G p.K297E Chr.1: 45345985-45345985 on Build GRCh38
3997548 c.896G>A p.R299H Chr.1: 45345930-45345930 on Build GRCh38
6800013 c.906delT p.F302fs*12 Chr.1: 45345968-45345968 on Build GRCh38
6800014 c.858delT p.F286fs*12 Chr.1: 45345968-45345968 on Build GRCh38
4008505 c.895C>T p.R299C Chr.1: 45345931-45345931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764501788 Chr.1: 45346001-45346001 on Build GRCh38
rs775045482 Chr.1: 45345916-45345916 on Build GRCh38
rs540919061 Chr.1: 45345967-45345967 on Build GRCh38
rs766290408 Chr.1: 45346036-45346036 on Build GRCh38
rs201509765 Chr.1: 45345891-45345891 on Build GRCh38
rs1052080683 Chr.1: 45345850-45345850 on Build GRCh38
rs761480948 Chr.1: 45345896-45345896 on Build GRCh38
rs192298497 Chr.1: 45345991-45345991 on Build GRCh38
rs369677376 Chr.1: 45345941-45345941 on Build GRCh38
rs747780572 Chr.1: 45345976-45345976 on Build GRCh38
rs911701591 Chr.1: 45345914-45345914 on Build GRCh38
rs776882268 Chr.1: 45346010-45346010 on Build GRCh38
rs769414518 Chr.1: 45345911-45345911 on Build GRCh38
rs202078785 Chr.1: 45345931-45345931 on Build GRCh38
rs778883567 Chr.1: 45345878-45345878 on Build GRCh38
rs371488726 Chr.1: 45345876-45345876 on Build GRCh38
rs750512795 Chr.1: 45346025-45346025 on Build GRCh38
rs753002813 Chr.1: 45345845-45345845 on Build GRCh38
rs201995698 Chr.1: 45345888-45345888 on Build GRCh38
rs758404137 Chr.1: 45346027-45346027 on Build GRCh38
rs780678665 Chr.1: 45345879-45345879 on Build GRCh38
rs766994146 Chr.1: 45345830-45345830 on Build GRCh38
rs752143450 Chr.1: 45346005-45346006 on Build GRCh38
rs558510789 Chr.1: 45345930-45345930 on Build GRCh38
rs767969691 Chr.1: 45345837-45345837 on Build GRCh38
rs769467396 Chr.1: 45345977-45345977 on Build GRCh38
rs1030357084 Chr.1: 45345973-45345973 on Build GRCh38
rs776277427 Chr.1: 45345892-45345892 on Build GRCh38
rs368668768 Chr.1: 45345866-45345866 on Build GRCh38
rs753018859 Chr.1: 45345926-45345926 on Build GRCh38
rs1056123497 Chr.1: 45345997-45345997 on Build GRCh38
rs755652208 Chr.1: 45346008-45346009 on Build GRCh38
rs760088342 Chr.1: 45345924-45345924 on Build GRCh38
rs754794946 Chr.1: 45345965-45345965 on Build GRCh38
rs758350727 Chr.1: 45345960-45345960 on Build GRCh38
rs771896778 Chr.1: 45345884-45345884 on Build GRCh38
rs758695163 Chr.1: 45345869-45345870 on Build GRCh38
rs376690303 Chr.1: 45345890-45345890 on Build GRCh38
rs748751819 Chr.1: 45345979-45345979 on Build GRCh38
rs760035503 Chr.1: 45345834-45345834 on Build GRCh38
rs761324690 Chr.1: 45345827-45345827 on Build GRCh38
rs369960423 Chr.1: 45346003-45346003 on Build GRCh38
rs774581306 Chr.1: 45345958-45345958 on Build GRCh38
rs35050933 Chr.1: 45345971-45345972 on Build GRCh38
rs964520189 Chr.1: 45345922-45345922 on Build GRCh38
rs201222735 Chr.1: 45345978-45345978 on Build GRCh38
rs370363350 Chr.1: 45345999-45345999 on Build GRCh38
rs953485326 Chr.1: 45345974-45345974 on Build GRCh38
rs772464710 Chr.1: 45345984-45345984 on Build GRCh38
rs753867300 Chr.1: 45345872-45345872 on Build GRCh38
rs187399031 Chr.1: 45345944-45345944 on Build GRCh38
rs1002024865 Chr.1: 45345921-45345921 on Build GRCh38
rs762055990 Chr.1: 45346013-45346013 on Build GRCh38
rs766859858 Chr.1: 45345911-45345913 on Build GRCh38
rs779980860 Chr.1: 45345962-45345962 on Build GRCh38
rs750866636 Chr.1: 45345846-45345847 on Build GRCh38
rs765397500 Chr.1: 45346023-45346023 on Build GRCh38
rs1023626425 Chr.1: 45345852-45345852 on Build GRCh38
rs757381218 Chr.1: 45345933-45345933 on Build GRCh38
rs756314558 Chr.1: 45345856-45345856 on Build GRCh38
rs752106801 Chr.1: 45345832-45345832 on Build GRCh38
rs761034307 Chr.1: 45345928-45345928 on Build GRCh38
rs372157706 Chr.1: 45345925-45345925 on Build GRCh38

View Full Product Details