Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53787091-53787336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDC1
Entrez Gene ID
55706
Gene Name
NDC1 transmembrane nucleoporin
Gene Aliases
NET3, TMEM48
UniGene
Hs.476525
Gene Chromosome Location
Chr.1: 53765460-53838552 on Build GRCh38
Transcripts
NM_001168551.1 NM_018087.4 NR_033142.1 XM_011541766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5352529 c.1700-6delC p.? Chr.1: 53787264-53787264 on Build GRCh38
6126930 c.1713A>T p.L571F Chr.1: 53787245-53787245 on Build GRCh38
1627122 c.1700-9delT p.? Chr.1: 53787267-53787267 on Build GRCh38
6723464 c.1757C>T p.T586M Chr.1: 53787201-53787201 on Build GRCh38
5483915 c.1794G>A p.L598L Chr.1: 53787164-53787164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368850743 Chr.1: 53787131-53787131 on Build GRCh38
rs572879190 Chr.1: 53787134-53787134 on Build GRCh38
rs750445957 Chr.1: 53787269-53787269 on Build GRCh38
rs763930653 Chr.1: 53787239-53787239 on Build GRCh38
rs187459137 Chr.1: 53787193-53787193 on Build GRCh38
rs758221560 Chr.1: 53787120-53787120 on Build GRCh38
rs376291549 Chr.1: 53787126-53787126 on Build GRCh38
rs765466131 Chr.1: 53787166-53787166 on Build GRCh38
rs762612478 Chr.1: 53787232-53787232 on Build GRCh38
rs374521168 Chr.1: 53787143-53787143 on Build GRCh38
rs751200195 Chr.1: 53787266-53787267 on Build GRCh38
rs751398108 Chr.1: 53787242-53787242 on Build GRCh38
rs530767688 Chr.1: 53787251-53787251 on Build GRCh38
rs140071660 Chr.1: 53787201-53787201 on Build GRCh38
rs780900051 Chr.1: 53787283-53787283 on Build GRCh38
rs544059155 Chr.1: 53787292-53787292 on Build GRCh38
rs558566839 Chr.1: 53787200-53787200 on Build GRCh38
rs749549884 Chr.1: 53787142-53787142 on Build GRCh38
rs917110808 Chr.1: 53787281-53787281 on Build GRCh38
rs142123604 Chr.1: 53787174-53787174 on Build GRCh38
rs202006036 Chr.1: 53787127-53787127 on Build GRCh38
rs780625392 Chr.1: 53787137-53787137 on Build GRCh38
rs762519783 Chr.1: 53787267-53787268 on Build GRCh38
rs367642403 Chr.1: 53787148-53787148 on Build GRCh38
rs764153102 Chr.1: 53787125-53787125 on Build GRCh38
rs575542532 Chr.1: 53787267-53787267 on Build GRCh38
rs756222543 Chr.1: 53787274-53787274 on Build GRCh38
rs754763312 Chr.1: 53787267-53787269 on Build GRCh38
rs61774803 Chr.1: 53787264-53787264 on Build GRCh38
rs867540922 Chr.1: 53787275-53787275 on Build GRCh38
rs370214243 Chr.1: 53787267-53787267 on Build GRCh38
rs985735319 Chr.1: 53787187-53787187 on Build GRCh38
rs1012821297 Chr.1: 53787114-53787114 on Build GRCh38
rs759515575 Chr.1: 53787116-53787116 on Build GRCh38
rs775500548 Chr.1: 53787222-53787222 on Build GRCh38
rs1047777008 Chr.1: 53787123-53787123 on Build GRCh38

View Full Product Details