Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39080814-39081077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3G
Entrez Gene ID
60489
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3G
Gene Aliases
A3G, ARCD, ARP-9, ARP9, CEM-15, CEM15, MDS019, bK150C2.7, dJ494G10.1
UniGene
Hs.660143
Gene Chromosome Location
Chr.22: 39077005-39087743 on Build GRCh38
Transcripts
NM_021822.3 XM_006724290.1 XM_017028903.1 XM_017028904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4818186 c.234G>T p.R78S Chr.22: 39080995-39080995 on Build GRCh38
1416308 c.291C>T p.C97C Chr.22: 39081052-39081052 on Build GRCh38
4818255 c.259G>A p.E87K Chr.22: 39081020-39081020 on Build GRCh38
5607795 c.213C>T p.F71F Chr.22: 39080974-39080974 on Build GRCh38
6602387 c.264C>T p.V88V Chr.22: 39081025-39081025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749736690 Chr.22: 39081002-39081002 on Build GRCh38
rs966846775 Chr.22: 39081044-39081044 on Build GRCh38
rs780221227 Chr.22: 39080959-39080959 on Build GRCh38
rs758427338 Chr.22: 39080950-39080950 on Build GRCh38
rs763628788 Chr.22: 39081007-39081007 on Build GRCh38
rs771432568 Chr.22: 39081003-39081003 on Build GRCh38
rs967653451 Chr.22: 39080960-39080960 on Build GRCh38
rs770182070 Chr.22: 39080895-39080895 on Build GRCh38
rs542618397 Chr.22: 39081014-39081014 on Build GRCh38
rs750252144 Chr.22: 39081043-39081043 on Build GRCh38
rs760249189 Chr.22: 39080918-39080918 on Build GRCh38
rs761071741 Chr.22: 39081018-39081018 on Build GRCh38
rs544919409 Chr.22: 39080974-39080974 on Build GRCh38
rs773578165 Chr.22: 39080993-39080993 on Build GRCh38
rs753523943 Chr.22: 39080935-39080935 on Build GRCh38
rs767071392 Chr.22: 39080906-39080906 on Build GRCh38
rs540940342 Chr.22: 39080876-39080876 on Build GRCh38
rs752216597 Chr.22: 39080912-39080912 on Build GRCh38
rs765028146 Chr.22: 39081035-39081035 on Build GRCh38
rs560885154 Chr.22: 39081050-39081050 on Build GRCh38
rs142924244 Chr.22: 39081005-39081005 on Build GRCh38
rs758375639 Chr.22: 39081049-39081049 on Build GRCh38
rs900959809 Chr.22: 39080947-39080947 on Build GRCh38
rs1011553055 Chr.22: 39080942-39080942 on Build GRCh38
rs371359635 Chr.22: 39080969-39080969 on Build GRCh38
rs763834861 Chr.22: 39080928-39080928 on Build GRCh38
rs761502799 Chr.22: 39081016-39081016 on Build GRCh38
rs201215107 Chr.22: 39080977-39080977 on Build GRCh38
rs747116946 Chr.22: 39080961-39080961 on Build GRCh38
rs773807793 Chr.22: 39080897-39080897 on Build GRCh38
rs1035901502 Chr.22: 39081001-39081001 on Build GRCh38
rs757122997 Chr.22: 39080939-39080939 on Build GRCh38
rs138285438 Chr.22: 39080975-39080975 on Build GRCh38
rs1048275662 Chr.22: 39080857-39080857 on Build GRCh38
rs577767262 Chr.22: 39080971-39080971 on Build GRCh38
rs769018026 Chr.22: 39080864-39080864 on Build GRCh38
rs760196037 Chr.22: 39081006-39081006 on Build GRCh38
rs112603901 Chr.22: 39080941-39080941 on Build GRCh38

View Full Product Details