Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30153591-30153864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM10
Entrez Gene ID
10107
Gene Name
tripartite motif containing 10
Gene Aliases
HERF1, RFB30, RNF9
UniGene
Hs.709483
Gene Chromosome Location
Chr.6: 30151939-30163216 on Build GRCh38
Transcripts
NM_006778.3 NM_052828.2 XM_011514221.1 XM_011514222.2 XM_011514223.2 XM_011514225.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5047437 c.1174G>A p.E392K Chr.6: 30153702-30153702 on Build GRCh38
4393584 c.1137C>T p.C379C Chr.6: 30153739-30153739 on Build GRCh38
2152082 c.1124G>A p.G375E Chr.6: 30153752-30153752 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750731963 Chr.6: 30153761-30153761 on Build GRCh38
rs765208131 Chr.6: 30153780-30153781 on Build GRCh38
rs577316825 Chr.6: 30153646-30153646 on Build GRCh38
rs759538414 Chr.6: 30153746-30153746 on Build GRCh38
rs12210298 Chr.6: 30153748-30153748 on Build GRCh38
rs9468694 Chr.6: 30153688-30153688 on Build GRCh38
rs992700998 Chr.6: 30153717-30153717 on Build GRCh38
rs561684446 Chr.6: 30153806-30153806 on Build GRCh38
rs774588443 Chr.6: 30153714-30153714 on Build GRCh38
rs570523320 Chr.6: 30153833-30153833 on Build GRCh38
rs148969734 Chr.6: 30153699-30153699 on Build GRCh38
rs780485179 Chr.6: 30153809-30153809 on Build GRCh38
rs751191960 Chr.6: 30153781-30153781 on Build GRCh38
rs770148993 Chr.6: 30153807-30153807 on Build GRCh38
rs766169289 Chr.6: 30153736-30153736 on Build GRCh38
rs878973550 Chr.6: 30153695-30153695 on Build GRCh38
rs915753690 Chr.6: 30153718-30153718 on Build GRCh38
rs771284773 Chr.6: 30153671-30153671 on Build GRCh38
rs987461314 Chr.6: 30153743-30153743 on Build GRCh38
rs920713194 Chr.6: 30153782-30153782 on Build GRCh38
rs775545025 Chr.6: 30153689-30153689 on Build GRCh38
rs546198554 Chr.6: 30153670-30153670 on Build GRCh38
rs753742225 Chr.6: 30153740-30153740 on Build GRCh38
rs147954414 Chr.6: 30153810-30153810 on Build GRCh38
rs781525167 Chr.6: 30153792-30153792 on Build GRCh38
rs376618536 Chr.6: 30153678-30153678 on Build GRCh38
rs3734840 Chr.6: 30153828-30153828 on Build GRCh38
rs373314409 Chr.6: 30153659-30153659 on Build GRCh38
rs756907085 Chr.6: 30153791-30153791 on Build GRCh38
rs761975346 Chr.6: 30153715-30153715 on Build GRCh38
rs2517649 Chr.6: 30153837-30153837 on Build GRCh38
rs374872920 Chr.6: 30153779-30153779 on Build GRCh38
rs376605686 Chr.6: 30153836-30153836 on Build GRCh38
rs752691257 Chr.6: 30153756-30153756 on Build GRCh38
rs757876056 Chr.6: 30153778-30153778 on Build GRCh38
rs768636858 Chr.6: 30153702-30153702 on Build GRCh38
rs748386559 Chr.6: 30153813-30153813 on Build GRCh38
rs772485615 Chr.6: 30153817-30153817 on Build GRCh38
rs1029227867 Chr.6: 30153687-30153687 on Build GRCh38
rs768744838 Chr.6: 30153811-30153811 on Build GRCh38
rs774336839 Chr.6: 30153812-30153812 on Build GRCh38
rs553187271 Chr.6: 30153727-30153727 on Build GRCh38
rs55670497 Chr.6: 30153834-30153834 on Build GRCh38
rs777425754 Chr.6: 30153655-30153655 on Build GRCh38
rs2022065 Chr.6: 30153683-30153683 on Build GRCh38
rs768564482 Chr.6: 30153825-30153825 on Build GRCh38

View Full Product Details