Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 39744271-39744522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THUMPD2
Entrez Gene ID
80745
Gene Name
THUMP domain containing 2
Gene Aliases
C2orf8
UniGene
Hs.468254
Gene Chromosome Location
Chr.2: 39736060-39779276 on Build GRCh38
Transcripts
NM_001321468.1 NM_001321469.1 NM_001321470.1 NM_001321474.1 NM_001321475.1 NM_001321477.1 NM_001321478.1 NM_001321479.1 NM_001321480.1 NM_001321481.1 NM_025264.4 NR_028102.1 NR_135659.1 NR_135660.1 NR_135661.1 NR_135662.1 NR_135663.1 NR_135664.1 XM_011533119.2 XM_017005050.1 XM_017005051.1 XM_017005054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
721000 c.1087G>T p.E363* Chr.2: 39744380-39744380 on Build GRCh38
5040767 c.1013T>G p.V338G Chr.2: 39744454-39744454 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764341541 Chr.2: 39744412-39744413 on Build GRCh38
rs750883034 Chr.2: 39744476-39744476 on Build GRCh38
rs776442089 Chr.2: 39744460-39744460 on Build GRCh38
rs770137041 Chr.2: 39744396-39744396 on Build GRCh38
rs764913285 Chr.2: 39744487-39744487 on Build GRCh38
rs754038978 Chr.2: 39744441-39744443 on Build GRCh38
rs772182382 Chr.2: 39744435-39744435 on Build GRCh38
rs914966150 Chr.2: 39744317-39744317 on Build GRCh38
rs536049570 Chr.2: 39744407-39744407 on Build GRCh38
rs151108984 Chr.2: 39744390-39744390 on Build GRCh38
rs943226657 Chr.2: 39744415-39744415 on Build GRCh38
rs753441818 Chr.2: 39744329-39744329 on Build GRCh38
rs774322283 Chr.2: 39744410-39744410 on Build GRCh38
rs74617165 Chr.2: 39744318-39744318 on Build GRCh38
rs760626063 Chr.2: 39744457-39744457 on Build GRCh38
rs989249434 Chr.2: 39744311-39744311 on Build GRCh38
rs986014138 Chr.2: 39744353-39744353 on Build GRCh38
rs761403018 Chr.2: 39744466-39744466 on Build GRCh38
rs775567243 Chr.2: 39744448-39744448 on Build GRCh38
rs749909393 Chr.2: 39744369-39744369 on Build GRCh38
rs978122022 Chr.2: 39744343-39744343 on Build GRCh38
rs756879572 Chr.2: 39744336-39744336 on Build GRCh38
rs550446469 Chr.2: 39744491-39744491 on Build GRCh38
rs755521596 Chr.2: 39744370-39744370 on Build GRCh38
rs911517483 Chr.2: 39744358-39744358 on Build GRCh38
rs760536232 Chr.2: 39744327-39744327 on Build GRCh38
rs763998514 Chr.2: 39744458-39744458 on Build GRCh38
rs369624208 Chr.2: 39744366-39744366 on Build GRCh38
rs199946304 Chr.2: 39744386-39744386 on Build GRCh38
rs749484381 Chr.2: 39744399-39744399 on Build GRCh38
rs531948150 Chr.2: 39744420-39744420 on Build GRCh38
rs763908726 Chr.2: 39744328-39744328 on Build GRCh38
rs574947314 Chr.2: 39744371-39744371 on Build GRCh38

View Full Product Details