Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54986040-54986295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM61
Entrez Gene ID
199964
Gene Name
transmembrane protein 61
Gene Aliases
-
UniGene
Hs.663950
Gene Chromosome Location
Chr.1: 54980614-54992293 on Build GRCh38
Transcripts
NM_182532.2 XM_005270586.4 XM_011540911.2 XM_011540912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2194265 c.25G>A p.G9R Chr.1: 54986106-54986106 on Build GRCh38
2194266 c.105C>T p.F35F Chr.1: 54986186-54986186 on Build GRCh38
1320983 c.151G>T p.A51S Chr.1: 54986232-54986232 on Build GRCh38
7394316 c.161C>T p.T54M Chr.1: 54986242-54986242 on Build GRCh38
4734575 c.188_189insC p.L65fs*64 Chr.1: 54986269-54986270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377660069 Chr.1: 54986131-54986131 on Build GRCh38
rs371535334 Chr.1: 54986252-54986252 on Build GRCh38
rs771903210 Chr.1: 54986105-54986105 on Build GRCh38
rs775542202 Chr.1: 54986251-54986251 on Build GRCh38
rs748897368 Chr.1: 54986170-54986170 on Build GRCh38
rs3737832 Chr.1: 54986199-54986199 on Build GRCh38
rs775008524 Chr.1: 54986262-54986262 on Build GRCh38
rs769714257 Chr.1: 54986178-54986178 on Build GRCh38
rs577669886 Chr.1: 54986171-54986171 on Build GRCh38
rs922467669 Chr.1: 54986243-54986243 on Build GRCh38
rs753149844 Chr.1: 54986134-54986134 on Build GRCh38
rs371703533 Chr.1: 54986111-54986111 on Build GRCh38
rs759637613 Chr.1: 54986130-54986130 on Build GRCh38
rs961680237 Chr.1: 54986223-54986223 on Build GRCh38
rs770666105 Chr.1: 54986180-54986180 on Build GRCh38
rs768239296 Chr.1: 54986090-54986090 on Build GRCh38
rs572352844 Chr.1: 54986113-54986113 on Build GRCh38
rs144225985 Chr.1: 54986187-54986187 on Build GRCh38
rs746839652 Chr.1: 54986179-54986179 on Build GRCh38
rs748057353 Chr.1: 54986101-54986101 on Build GRCh38
rs778412005 Chr.1: 54986213-54986213 on Build GRCh38
rs752016944 Chr.1: 54986157-54986157 on Build GRCh38
rs755887453 Chr.1: 54986079-54986079 on Build GRCh38
rs1026691784 Chr.1: 54986268-54986268 on Build GRCh38
rs764092454 Chr.1: 54986082-54986084 on Build GRCh38
rs767930980 Chr.1: 54986204-54986204 on Build GRCh38
rs181322664 Chr.1: 54986151-54986151 on Build GRCh38
rs772100194 Chr.1: 54986176-54986176 on Build GRCh38
rs201343395 Chr.1: 54986198-54986198 on Build GRCh38
rs754721430 Chr.1: 54986212-54986212 on Build GRCh38
rs768565621 Chr.1: 54986255-54986255 on Build GRCh38
rs139250137 Chr.1: 54986182-54986182 on Build GRCh38
rs757173118 Chr.1: 54986192-54986193 on Build GRCh38
rs781032962 Chr.1: 54986238-54986238 on Build GRCh38
rs753660181 Chr.1: 54986188-54986189 on Build GRCh38
rs777711469 Chr.1: 54986229-54986229 on Build GRCh38
rs147533721 Chr.1: 54986120-54986120 on Build GRCh38
rs768131027 Chr.1: 54986274-54986274 on Build GRCh38
rs563563686 Chr.1: 54986106-54986106 on Build GRCh38
rs138273726 Chr.1: 54986242-54986242 on Build GRCh38
rs779829125 Chr.1: 54986086-54986086 on Build GRCh38
rs757809970 Chr.1: 54986225-54986225 on Build GRCh38
rs753541269 Chr.1: 54986211-54986211 on Build GRCh38
rs765371477 Chr.1: 54986192-54986192 on Build GRCh38
rs759010696 Chr.1: 54986150-54986150 on Build GRCh38
rs762341441 Chr.1: 54986270-54986270 on Build GRCh38
rs747602597 Chr.1: 54986172-54986172 on Build GRCh38
rs145543120 Chr.1: 54986234-54986234 on Build GRCh38
rs762288382 Chr.1: 54986201-54986201 on Build GRCh38
rs148773188 Chr.1: 54986177-54986177 on Build GRCh38
rs748844087 Chr.1: 54986087-54986087 on Build GRCh38
rs779880277 Chr.1: 54986169-54986169 on Build GRCh38
rs370940522 Chr.1: 54986193-54986193 on Build GRCh38
rs144119137 Chr.1: 54986160-54986160 on Build GRCh38
rs200980364 Chr.1: 54986214-54986214 on Build GRCh38
rs750869581 Chr.1: 54986276-54986276 on Build GRCh38
rs143466854 Chr.1: 54986097-54986097 on Build GRCh38
rs760344355 Chr.1: 54986072-54986081 on Build GRCh38

View Full Product Details