Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53258776-53259025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VN1R2
Entrez Gene ID
317701
Gene Name
vomeronasal 1 receptor 2
Gene Aliases
V1RL2
UniGene
Hs.553684
Gene Chromosome Location
Chr.19: 53258292-53259602 on Build GRCh38
Transcripts
NM_173856.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3538326 c.650G>A p.W217* Chr.19: 53259025-53259025 on Build GRCh38
1396082 c.598G>T p.E200* Chr.19: 53258973-53258973 on Build GRCh38
3538325 c.493C>T p.L165F Chr.19: 53258868-53258868 on Build GRCh38
1396080 c.536G>A p.S179N Chr.19: 53258911-53258911 on Build GRCh38
130158 c.530T>A p.I177N Chr.19: 53258905-53258905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368744499 Chr.19: 53258871-53258871 on Build GRCh38
rs200295676 Chr.19: 53258905-53258905 on Build GRCh38
rs149739314 Chr.19: 53258996-53258996 on Build GRCh38
rs374405049 Chr.19: 53258968-53258968 on Build GRCh38
rs1008192407 Chr.19: 53258930-53258930 on Build GRCh38
rs775171130 Chr.19: 53258807-53258807 on Build GRCh38
rs776291171 Chr.19: 53258928-53258928 on Build GRCh38
rs753939442 Chr.19: 53258977-53258977 on Build GRCh38
rs770224742 Chr.19: 53258865-53258865 on Build GRCh38
rs760146458 Chr.19: 53258823-53258823 on Build GRCh38
rs375291938 Chr.19: 53258971-53258971 on Build GRCh38
rs771662065 Chr.19: 53258938-53258938 on Build GRCh38
rs771150853 Chr.19: 53258923-53258923 on Build GRCh38
rs925684777 Chr.19: 53258846-53258846 on Build GRCh38
rs767366544 Chr.19: 53258921-53258921 on Build GRCh38
rs61751885 Chr.19: 53258844-53258844 on Build GRCh38
rs369017056 Chr.19: 53258993-53258993 on Build GRCh38
rs781715655 Chr.19: 53258834-53258834 on Build GRCh38
rs147886409 Chr.19: 53258921-53258922 on Build GRCh38
rs768122096 Chr.19: 53258926-53258926 on Build GRCh38
rs745683925 Chr.19: 53258904-53258904 on Build GRCh38
rs201557508 Chr.19: 53258934-53258934 on Build GRCh38
rs114416919 Chr.19: 53258883-53258883 on Build GRCh38
rs778865222 Chr.19: 53258992-53258992 on Build GRCh38
rs774965789 Chr.19: 53258942-53258942 on Build GRCh38
rs371881645 Chr.19: 53258875-53258875 on Build GRCh38
rs773919141 Chr.19: 53258831-53258831 on Build GRCh38
rs768194944 Chr.19: 53258828-53258828 on Build GRCh38
rs775249431 Chr.19: 53258944-53258944 on Build GRCh38
rs779625161 Chr.19: 53258913-53258913 on Build GRCh38
rs201211389 Chr.19: 53258896-53258896 on Build GRCh38
rs746375950 Chr.19: 53258862-53258862 on Build GRCh38
rs759346113 Chr.19: 53258915-53258915 on Build GRCh38
rs751992274 Chr.19: 53258863-53258863 on Build GRCh38
rs774213336 Chr.19: 53258851-53258851 on Build GRCh38
rs544930366 Chr.19: 53258999-53258999 on Build GRCh38
rs763555184 Chr.19: 53258834-53258834 on Build GRCh38
rs534711649 Chr.19: 53258870-53258870 on Build GRCh38
rs775851635 Chr.19: 53258893-53258893 on Build GRCh38
rs867608921 Chr.19: 53258895-53258895 on Build GRCh38
rs752929083 Chr.19: 53258952-53258952 on Build GRCh38
rs1018378963 Chr.19: 53258937-53258937 on Build GRCh38
rs757349017 Chr.19: 53258818-53258818 on Build GRCh38
rs1047748993 Chr.19: 53258852-53258852 on Build GRCh38
rs771593869 Chr.19: 53258941-53258941 on Build GRCh38
rs756206322 Chr.19: 53258878-53258878 on Build GRCh38
rs767910840 Chr.19: 53258951-53258951 on Build GRCh38
rs577019316 Chr.19: 53258845-53258845 on Build GRCh38
rs757289820 Chr.19: 53258987-53258987 on Build GRCh38

View Full Product Details