Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10908681-10908942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRH1
Entrez Gene ID
5554
Gene Name
proline rich protein HaeIII subfamily 1
Gene Aliases
Db-s, PA, PIF-S, PRH2, Pr1/Pr2
UniGene
Hs.656965
Gene Chromosome Location
Chr.12: 10880961-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_037918.2 NM_023920.2

Gene Symbol
PRH1-PRR4
Entrez Gene ID
100533464
Gene Name
PRH1-PRR4 readthrough
Gene Aliases
PRH1
UniGene
Hs.732031
Gene Chromosome Location
Chr.12: 10845849-11171625 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_037918.2 NM_023920.2

Gene Symbol
TAS2R13
Entrez Gene ID
50838
Gene Name
taste 2 receptor member 13
Gene Aliases
T2R13, TRB3
UniGene
Hs.679407
Gene Chromosome Location
Chr.12: 10907926-10909562 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_037918.2 NM_023920.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2150088 c.411C>T p.T137T Chr.12: 10908888-10908888 on Build GRCh38
4186499 c.538A>C p.K180Q Chr.12: 10908761-10908761 on Build GRCh38
32592 c.446A>G p.N149S Chr.12: 10908853-10908853 on Build GRCh38
109624 c.612C>T p.S204S Chr.12: 10908687-10908687 on Build GRCh38
6567825 c.537C>G p.V179V Chr.12: 10908762-10908762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763264582 Chr.12: 10908783-10908783 on Build GRCh38
rs768082133 Chr.12: 10908716-10908716 on Build GRCh38
rs781172305 Chr.12: 10908760-10908760 on Build GRCh38
rs774795106 Chr.12: 10908811-10908811 on Build GRCh38
rs762206371 Chr.12: 10908812-10908812 on Build GRCh38
rs751826901 Chr.12: 10908708-10908708 on Build GRCh38
rs376987737 Chr.12: 10908808-10908808 on Build GRCh38
rs775398971 Chr.12: 10908769-10908769 on Build GRCh38
rs894927278 Chr.12: 10908819-10908819 on Build GRCh38
rs752444755 Chr.12: 10908751-10908751 on Build GRCh38
rs375488942 Chr.12: 10908721-10908721 on Build GRCh38
rs145310730 Chr.12: 10908869-10908869 on Build GRCh38
rs183634674 Chr.12: 10908862-10908862 on Build GRCh38
rs780202524 Chr.12: 10908899-10908899 on Build GRCh38
rs200842780 Chr.12: 10908746-10908746 on Build GRCh38
rs372765078 Chr.12: 10908784-10908784 on Build GRCh38
rs769906007 Chr.12: 10908766-10908766 on Build GRCh38
rs141663427 Chr.12: 10908759-10908759 on Build GRCh38
rs762435629 Chr.12: 10908713-10908713 on Build GRCh38
rs754606395 Chr.12: 10908744-10908744 on Build GRCh38
rs368652295 Chr.12: 10908758-10908758 on Build GRCh38
rs148234617 Chr.12: 10908752-10908752 on Build GRCh38
rs777222102 Chr.12: 10908853-10908853 on Build GRCh38
rs761242032 Chr.12: 10908836-10908836 on Build GRCh38
rs753408054 Chr.12: 10908737-10908737 on Build GRCh38
rs769675563 Chr.12: 10908897-10908897 on Build GRCh38
rs975168806 Chr.12: 10908778-10908778 on Build GRCh38
rs777657052 Chr.12: 10908753-10908753 on Build GRCh38
rs142394171 Chr.12: 10908908-10908908 on Build GRCh38
rs764557255 Chr.12: 10908705-10908705 on Build GRCh38
rs200499053 Chr.12: 10908861-10908861 on Build GRCh38
rs745787811 Chr.12: 10908765-10908765 on Build GRCh38
rs780938834 Chr.12: 10908867-10908867 on Build GRCh38
rs767994152 Chr.12: 10908822-10908822 on Build GRCh38
rs751029522 Chr.12: 10908829-10908829 on Build GRCh38
rs137927359 Chr.12: 10908882-10908882 on Build GRCh38
rs752111217 Chr.12: 10908850-10908850 on Build GRCh38
rs764844013 Chr.12: 10908847-10908847 on Build GRCh38
rs902722400 Chr.12: 10908833-10908833 on Build GRCh38
rs570122150 Chr.12: 10908814-10908814 on Build GRCh38
rs749272501 Chr.12: 10908903-10908903 on Build GRCh38
rs746721818 Chr.12: 10908754-10908754 on Build GRCh38

View Full Product Details