Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 637602-637829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP72
Entrez Gene ID
55722
Gene Name
centrosomal protein 72
Gene Aliases
-
UniGene
Hs.591741
Gene Chromosome Location
Chr.5: 612290-663500 on Build GRCh38
Transcripts
NM_018140.3 XM_005248322.3 XM_011514063.1 XM_011514064.2 XM_017009626.1 XM_017009627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855856 c.1058G>A p.C353Y Chr.5: 637670-637670 on Build GRCh38
3855855 c.1031C>T p.T344M Chr.5: 637643-637643 on Build GRCh38
4522271 c.1153G>C p.E385Q Chr.5: 637765-637765 on Build GRCh38
1069304 c.1004G>A p.R335Q Chr.5: 637616-637616 on Build GRCh38
3345888 c.1074T>G p.H358Q Chr.5: 637686-637686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780488743 Chr.5: 637720-637720 on Build GRCh38
rs267600673 Chr.5: 637774-637774 on Build GRCh38
rs896688381 Chr.5: 637701-637701 on Build GRCh38
rs748920540 Chr.5: 637671-637671 on Build GRCh38
rs753929591 Chr.5: 637686-637686 on Build GRCh38
rs187718758 Chr.5: 637647-637647 on Build GRCh38
rs763789184 Chr.5: 637743-637743 on Build GRCh38
rs747715042 Chr.5: 637755-637755 on Build GRCh38
rs750368955 Chr.5: 637661-637661 on Build GRCh38
rs758738113 Chr.5: 637716-637716 on Build GRCh38
rs988957304 Chr.5: 637634-637634 on Build GRCh38
rs751266368 Chr.5: 637674-637674 on Build GRCh38
rs940552272 Chr.5: 637739-637739 on Build GRCh38
rs766190281 Chr.5: 637772-637772 on Build GRCh38
rs774203306 Chr.5: 637773-637773 on Build GRCh38
rs62001002 Chr.5: 637644-637644 on Build GRCh38
rs369296828 Chr.5: 637780-637780 on Build GRCh38
rs762974032 Chr.5: 637769-637769 on Build GRCh38
rs375668537 Chr.5: 637796-637796 on Build GRCh38
rs551556889 Chr.5: 637695-637695 on Build GRCh38
rs201002445 Chr.5: 637795-637795 on Build GRCh38
rs776362091 Chr.5: 637746-637746 on Build GRCh38
rs931717942 Chr.5: 637783-637783 on Build GRCh38
rs201362331 Chr.5: 637696-637696 on Build GRCh38
rs757460319 Chr.5: 637657-637657 on Build GRCh38
rs140824465 Chr.5: 637685-637685 on Build GRCh38
rs1013760633 Chr.5: 637725-637725 on Build GRCh38
rs750420955 Chr.5: 637807-637807 on Build GRCh38
rs766179054 Chr.5: 637673-637673 on Build GRCh38
rs747294322 Chr.5: 637729-637729 on Build GRCh38
rs994450589 Chr.5: 637650-637650 on Build GRCh38
rs775044934 Chr.5: 637642-637642 on Build GRCh38
rs765278663 Chr.5: 637806-637806 on Build GRCh38
rs143910441 Chr.5: 637649-637649 on Build GRCh38
rs770538271 Chr.5: 637768-637768 on Build GRCh38
rs762718833 Chr.5: 637671-637671 on Build GRCh38
rs757317781 Chr.5: 637693-637693 on Build GRCh38
rs980517276 Chr.5: 637653-637653 on Build GRCh38
rs760351806 Chr.5: 637643-637643 on Build GRCh38
rs1026300576 Chr.5: 637666-637666 on Build GRCh38
rs773076945 Chr.5: 637761-637761 on Build GRCh38
rs769420298 Chr.5: 637759-637759 on Build GRCh38
rs546473646 Chr.5: 637715-637715 on Build GRCh38
rs756393827 Chr.5: 637676-637676 on Build GRCh38
rs369797124 Chr.5: 637723-637723 on Build GRCh38
rs765276251 Chr.5: 637655-637655 on Build GRCh38

View Full Product Details