Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66261913-66262162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLC2
Entrez Gene ID
64837
Gene Name
kinesin light chain 2
Gene Aliases
-
UniGene
Hs.280792
Gene Chromosome Location
Chr.11: 66257294-66267861 on Build GRCh38
Transcripts
NM_001134774.1 NM_001134775.1 NM_001134776.1 NM_001318734.1 NM_022822.2 XM_005274183.1 XM_005274184.1 XM_017018133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4923286 c.460-2A>T p.? Chr.11: 66262121-66262121 on Build GRCh38
127138 c.459+1G>C p.? Chr.11: 66261973-66261973 on Build GRCh38
4496708 c.480C>T p.P160P Chr.11: 66262143-66262143 on Build GRCh38
930683 c.449C>T p.A150V Chr.11: 66261962-66261962 on Build GRCh38
253756 c.459C>T p.N153N Chr.11: 66261972-66261972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763803545 Chr.11: 66262022-66262022 on Build GRCh38
rs549150930 Chr.11: 66261944-66261944 on Build GRCh38
rs368665608 Chr.11: 66261980-66261980 on Build GRCh38
rs539851791 Chr.11: 66262073-66262073 on Build GRCh38
rs376235107 Chr.11: 66262011-66262011 on Build GRCh38
rs746187535 Chr.11: 66261966-66261966 on Build GRCh38
rs374310320 Chr.11: 66261947-66261947 on Build GRCh38
rs777110967 Chr.11: 66261972-66261973 on Build GRCh38
rs757760061 Chr.11: 66261960-66261960 on Build GRCh38
rs776453374 Chr.11: 66261986-66261986 on Build GRCh38
rs768931892 Chr.11: 66262108-66262108 on Build GRCh38
rs911167025 Chr.11: 66262103-66262103 on Build GRCh38
rs773382725 Chr.11: 66262138-66262138 on Build GRCh38
rs766360636 Chr.11: 66261983-66261983 on Build GRCh38
rs756690945 Chr.11: 66262089-66262089 on Build GRCh38
rs781669694 Chr.11: 66261961-66261961 on Build GRCh38
rs369947993 Chr.11: 66262097-66262097 on Build GRCh38
rs998016734 Chr.11: 66262070-66262070 on Build GRCh38
rs776578635 Chr.11: 66262057-66262057 on Build GRCh38
rs990041669 Chr.11: 66262136-66262136 on Build GRCh38
rs764910995 Chr.11: 66262004-66262004 on Build GRCh38
rs1056069555 Chr.11: 66261952-66261952 on Build GRCh38
rs200705720 Chr.11: 66262119-66262119 on Build GRCh38
rs41302421 Chr.11: 66262019-66262019 on Build GRCh38
rs1032949550 Chr.11: 66262047-66262047 on Build GRCh38
rs758112769 Chr.11: 66261943-66261943 on Build GRCh38
rs143892317 Chr.11: 66262031-66262031 on Build GRCh38
rs1047438553 Chr.11: 66261955-66261955 on Build GRCh38
rs199947030 Chr.11: 66262117-66262117 on Build GRCh38
rs886194580 Chr.11: 66261958-66261958 on Build GRCh38
rs868457311 Chr.11: 66262137-66262137 on Build GRCh38
rs769052713 Chr.11: 66261978-66261978 on Build GRCh38
rs748187904 Chr.11: 66262115-66262115 on Build GRCh38
rs749445604 Chr.11: 66261973-66261973 on Build GRCh38
rs770080117 Chr.11: 66261972-66261972 on Build GRCh38
rs1006410662 Chr.11: 66262029-66262029 on Build GRCh38
rs371920997 Chr.11: 66262001-66262001 on Build GRCh38
rs367882054 Chr.11: 66262075-66262075 on Build GRCh38
rs779291656 Chr.11: 66262109-66262109 on Build GRCh38
rs996120122 Chr.11: 66262037-66262037 on Build GRCh38
rs962186077 Chr.11: 66262054-66262054 on Build GRCh38
rs756776522 Chr.11: 66262074-66262074 on Build GRCh38
rs761987950 Chr.11: 66261981-66261981 on Build GRCh38
rs750860943 Chr.11: 66262078-66262078 on Build GRCh38

View Full Product Details