Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 91266673-91266908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD1
Entrez Gene ID
8321
Gene Name
frizzled class receptor 1
Gene Aliases
-
UniGene
Hs.94234
Gene Chromosome Location
Chr.7: 91264468-91268817 on Build GRCh38
Transcripts
NM_003505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3883262 c.1870G>A p.G624S Chr.7: 91266750-91266750 on Build GRCh38
6668096 c.1825A>T p.M609L Chr.7: 91266705-91266705 on Build GRCh38
3883260 c.1836C>T p.I612I Chr.7: 91266716-91266716 on Build GRCh38
6341313 c.1804G>T p.V602F Chr.7: 91266684-91266684 on Build GRCh38
3883261 c.1862T>A p.I621N Chr.7: 91266742-91266742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117293522 Chr.7: 91266763-91266763 on Build GRCh38
rs762285044 Chr.7: 91266855-91266871 on Build GRCh38
rs763445984 Chr.7: 91266716-91266716 on Build GRCh38
rs780333274 Chr.7: 91266781-91266781 on Build GRCh38
rs747130384 Chr.7: 91266791-91266791 on Build GRCh38
rs767160566 Chr.7: 91266830-91266830 on Build GRCh38
rs749290137 Chr.7: 91266796-91266796 on Build GRCh38
rs534296694 Chr.7: 91266825-91266825 on Build GRCh38
rs775212277 Chr.7: 91266839-91266839 on Build GRCh38
rs776800679 Chr.7: 91266792-91266792 on Build GRCh38
rs760207486 Chr.7: 91266846-91266846 on Build GRCh38
rs774358844 Chr.7: 91266828-91266828 on Build GRCh38
rs759395202 Chr.7: 91266829-91266829 on Build GRCh38
rs757714719 Chr.7: 91266742-91266742 on Build GRCh38
rs151338078 Chr.7: 91266719-91266719 on Build GRCh38
rs758605350 Chr.7: 91266776-91266776 on Build GRCh38
rs746224793 Chr.7: 91266770-91266770 on Build GRCh38
rs755665078 Chr.7: 91266710-91266710 on Build GRCh38
rs763684265 Chr.7: 91266847-91266847 on Build GRCh38
rs756059530 Chr.7: 91266765-91266765 on Build GRCh38
rs757825463 Chr.7: 91266852-91266852 on Build GRCh38
rs752211281 Chr.7: 91266709-91266709 on Build GRCh38
rs993880464 Chr.7: 91266795-91266795 on Build GRCh38
rs753312902 Chr.7: 91266848-91266848 on Build GRCh38

View Full Product Details