Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128462553-128462812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB8
Entrez Gene ID
165721
Gene Name
DnaJ heat shock protein family (Hsp40) member B8
Gene Aliases
CT156, DJ6
UniGene
Hs.518241
Gene Chromosome Location
Chr.3: 128462432-128467248 on Build GRCh38
Transcripts
NM_153330.4 XM_006713519.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
239620 c.619G>A p.V207M Chr.3: 128462627-128462627 on Build GRCh38
3940268 c.543G>A p.S181S Chr.3: 128462703-128462703 on Build GRCh38
6647155 c.598G>A p.V200M Chr.3: 128462648-128462648 on Build GRCh38
5111038 c.564G>A p.M188I Chr.3: 128462682-128462682 on Build GRCh38
3914854 c.669G>A p.K223K Chr.3: 128462577-128462577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2953129 Chr.3: 128462694-128462694 on Build GRCh38
rs935843145 Chr.3: 128462660-128462660 on Build GRCh38
rs1025782669 Chr.3: 128462752-128462752 on Build GRCh38
rs780012430 Chr.3: 128462742-128462742 on Build GRCh38
rs1039631559 Chr.3: 128462645-128462645 on Build GRCh38
rs199505596 Chr.3: 128462639-128462639 on Build GRCh38
rs754332024 Chr.3: 128462654-128462654 on Build GRCh38
rs778094237 Chr.3: 128462721-128462721 on Build GRCh38
rs748553089 Chr.3: 128462734-128462734 on Build GRCh38
rs758689838 Chr.3: 128462667-128462667 on Build GRCh38
rs774012058 Chr.3: 128462700-128462700 on Build GRCh38
rs529782278 Chr.3: 128462582-128462582 on Build GRCh38
rs118145329 Chr.3: 128462739-128462739 on Build GRCh38
rs765912735 Chr.3: 128462707-128462707 on Build GRCh38
rs757410955 Chr.3: 128462786-128462786 on Build GRCh38
rs776594226 Chr.3: 128462687-128462687 on Build GRCh38
rs770940516 Chr.3: 128462678-128462678 on Build GRCh38
rs770871823 Chr.3: 128462617-128462617 on Build GRCh38
rs865944606 Chr.3: 128462725-128462725 on Build GRCh38
rs748384137 Chr.3: 128462606-128462606 on Build GRCh38
rs1026507026 Chr.3: 128462751-128462751 on Build GRCh38
rs765817483 Chr.3: 128462773-128462773 on Build GRCh38
rs1038560766 Chr.3: 128462720-128462720 on Build GRCh38
rs36034445 Chr.3: 128462628-128462628 on Build GRCh38
rs374250309 Chr.3: 128462716-128462716 on Build GRCh38
rs759330041 Chr.3: 128462622-128462622 on Build GRCh38
rs757298895 Chr.3: 128462735-128462735 on Build GRCh38
rs779236523 Chr.3: 128462598-128462598 on Build GRCh38
rs144594699 Chr.3: 128462629-128462629 on Build GRCh38
rs756519699 Chr.3: 128462630-128462630 on Build GRCh38
rs747091190 Chr.3: 128462677-128462677 on Build GRCh38
rs760272118 Chr.3: 128462772-128462772 on Build GRCh38
rs768271693 Chr.3: 128462696-128462696 on Build GRCh38
rs764974602 Chr.3: 128462627-128462627 on Build GRCh38
rs755301106 Chr.3: 128462655-128462655 on Build GRCh38
rs140414363 Chr.3: 128462695-128462695 on Build GRCh38
rs749639687 Chr.3: 128462588-128462588 on Build GRCh38
rs751750410 Chr.3: 128462782-128462782 on Build GRCh38
rs750443437 Chr.3: 128462790-128462790 on Build GRCh38
rs35948511 Chr.3: 128462789-128462789 on Build GRCh38
rs772220976 Chr.3: 128462609-128462609 on Build GRCh38
rs915237255 Chr.3: 128462778-128462778 on Build GRCh38
rs750876122 Chr.3: 128462581-128462581 on Build GRCh38
rs370646538 Chr.3: 128462715-128462715 on Build GRCh38
rs199584005 Chr.3: 128462648-128462648 on Build GRCh38
rs147724414 Chr.3: 128462755-128462755 on Build GRCh38
rs747153425 Chr.3: 128462612-128462612 on Build GRCh38
rs200955342 Chr.3: 128462738-128462738 on Build GRCh38
rs148414049 Chr.3: 128462640-128462640 on Build GRCh38
rs1023222584 Chr.3: 128462723-128462723 on Build GRCh38
rs563290082 Chr.3: 128462688-128462688 on Build GRCh38
rs3749370 Chr.3: 128462704-128462704 on Build GRCh38
rs375861387 Chr.3: 128462774-128462774 on Build GRCh38
rs374581480 Chr.3: 128462749-128462749 on Build GRCh38
rs761344836 Chr.3: 128462702-128462702 on Build GRCh38
rs960848602 Chr.3: 128462757-128462757 on Build GRCh38
rs374525334 Chr.3: 128462610-128462610 on Build GRCh38
rs138648780 Chr.3: 128462583-128462583 on Build GRCh38
rs200292314 Chr.3: 128462596-128462596 on Build GRCh38
rs769577015 Chr.3: 128462693-128462693 on Build GRCh38
rs140895506 Chr.3: 128462775-128462775 on Build GRCh38
rs768468155 Chr.3: 128462746-128462746 on Build GRCh38
rs369582301 Chr.3: 128462666-128462666 on Build GRCh38
rs373509985 Chr.3: 128462674-128462674 on Build GRCh38
rs772593121 Chr.3: 128462767-128462767 on Build GRCh38
rs771814098 Chr.3: 128462761-128462761 on Build GRCh38
rs779433817 Chr.3: 128462661-128462661 on Build GRCh38
rs764560694 Chr.3: 128462776-128462776 on Build GRCh38
rs147231121 Chr.3: 128462703-128462703 on Build GRCh38
rs925384742 Chr.3: 128462653-128462653 on Build GRCh38
rs141628776 Chr.3: 128462649-128462649 on Build GRCh38

View Full Product Details