Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63338587-63338848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDSR
Entrez Gene ID
2531
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
DHSR, FVT1, SDR35C1
UniGene
Hs.74050
Gene Chromosome Location
Chr.18: 63327736-63367273 on Build GRCh38
Transcripts
NM_002035.2 XM_005266677.2 XM_017025690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
312239 c.730G>T p.V244L Chr.18: 63338847-63338847 on Build GRCh38
5686018 c.751G>T p.A251S Chr.18: 63338826-63338826 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs150958478 Chr.18: 63338639-63338639 on Build GRCh38
rs928287165 Chr.18: 63338660-63338660 on Build GRCh38
rs764767580 Chr.18: 63338817-63338820 on Build GRCh38
rs879343758 Chr.18: 63338698-63338698 on Build GRCh38
rs758319593 Chr.18: 63338754-63338754 on Build GRCh38
rs756349684 Chr.18: 63338800-63338800 on Build GRCh38
rs752707637 Chr.18: 63338751-63338751 on Build GRCh38
rs547011302 Chr.18: 63338777-63338777 on Build GRCh38
rs777472509 Chr.18: 63338757-63338757 on Build GRCh38
rs780320947 Chr.18: 63338802-63338802 on Build GRCh38
rs749446295 Chr.18: 63338803-63338803 on Build GRCh38
rs140763153 Chr.18: 63338707-63338707 on Build GRCh38
rs774470146 Chr.18: 63338741-63338741 on Build GRCh38
rs950417014 Chr.18: 63338683-63338683 on Build GRCh38
rs969434194 Chr.18: 63338755-63338755 on Build GRCh38
rs751349538 Chr.18: 63338764-63338764 on Build GRCh38
rs969753292 Chr.18: 63338638-63338638 on Build GRCh38

View Full Product Details