Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108734751-108734980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS4
Entrez Gene ID
8471
Gene Name
insulin receptor substrate 4
Gene Aliases
IRS-4, PY160
UniGene
Hs.407141
Gene Chromosome Location
Chr.X: 108719949-108736584 on Build GRCh38
Transcripts
NM_003604.2 XM_005262220.2 XM_006724713.3 XM_011531061.1

SNP Detail

SNP ID Chromosome Location
rs761223077 Chr.X: 108734869-108734869 on Build GRCh38
rs1037462222 Chr.X: 108734924-108734924 on Build GRCh38
rs778907991 Chr.X: 108734891-108734891 on Build GRCh38
rs775620395 Chr.X: 108734829-108734829 on Build GRCh38
rs373677981 Chr.X: 108734882-108734882 on Build GRCh38
rs755092846 Chr.X: 108734789-108734789 on Build GRCh38
rs762544135 Chr.X: 108734834-108734834 on Build GRCh38
rs777883481 Chr.X: 108734930-108734930 on Build GRCh38
rs369718541 Chr.X: 108734812-108734812 on Build GRCh38
rs749346215 Chr.X: 108734773-108734773 on Build GRCh38
rs928579393 Chr.X: 108734849-108734849 on Build GRCh38
rs779104385 Chr.X: 108734807-108734807 on Build GRCh38
rs758610761 Chr.X: 108734896-108734896 on Build GRCh38
rs888661272 Chr.X: 108734792-108734792 on Build GRCh38
rs774008589 Chr.X: 108734851-108734851 on Build GRCh38
rs146331588 Chr.X: 108734853-108734853 on Build GRCh38
rs138444058 Chr.X: 108734872-108734872 on Build GRCh38
rs767107481 Chr.X: 108734863-108734863 on Build GRCh38
rs748185194 Chr.X: 108734893-108734893 on Build GRCh38
rs780560532 Chr.X: 108734770-108734770 on Build GRCh38
rs745657679 Chr.X: 108734941-108734941 on Build GRCh38
rs748382230 Chr.X: 108734909-108734911 on Build GRCh38
rs769647116 Chr.X: 108734839-108734856 on Build GRCh38
rs17847225 Chr.X: 108734860-108734860 on Build GRCh38
rs34287560 Chr.X: 108734821-108734821 on Build GRCh38
rs748462108 Chr.X: 108734808-108734808 on Build GRCh38
rs769775639 Chr.X: 108734950-108734950 on Build GRCh38
rs769725218 Chr.X: 108734822-108734822 on Build GRCh38
rs1015888477 Chr.X: 108734850-108734850 on Build GRCh38
rs1030133699 Chr.X: 108734932-108734932 on Build GRCh38
rs763752721 Chr.X: 108734845-108734845 on Build GRCh38
rs200202171 Chr.X: 108734811-108734811 on Build GRCh38
rs750749821 Chr.X: 108734864-108734864 on Build GRCh38

View Full Product Details