Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37704846-37705117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNF1B
Entrez Gene ID
6928
Gene Name
HNF1 homeobox B
Gene Aliases
FJHN, HNF-1-beta, HNF-1B, HNF1beta, HNF2, HPC11, LF-B3, LFB3, MODY5, TCF-2, TCF2, VHNF1
UniGene
Hs.191144
Gene Chromosome Location
Chr.17: 37686431-37745078 on Build GRCh38
Transcripts
NM_000458.3 NM_001165923.3 NM_001304286.1 XM_011525160.1 XM_011525161.1 XM_011525164.1

SNP Detail

SNP ID Chromosome Location
rs952410476 Chr.17: 37705092-37705092 on Build GRCh38
rs201113814 Chr.17: 37705017-37705017 on Build GRCh38
rs371467412 Chr.17: 37705034-37705034 on Build GRCh38
rs748391570 Chr.17: 37704899-37704899 on Build GRCh38
rs1025261798 Chr.17: 37704987-37704987 on Build GRCh38
rs776536485 Chr.17: 37704918-37704918 on Build GRCh38
rs368068840 Chr.17: 37705054-37705054 on Build GRCh38
rs753417964 Chr.17: 37704888-37704888 on Build GRCh38
rs778319071 Chr.17: 37705019-37705019 on Build GRCh38
rs374854968 Chr.17: 37704900-37704900 on Build GRCh38
rs994368898 Chr.17: 37705074-37705074 on Build GRCh38
rs766076288 Chr.17: 37704981-37704981 on Build GRCh38
rs80237325 Chr.17: 37705000-37705000 on Build GRCh38
rs745872139 Chr.17: 37705048-37705048 on Build GRCh38
rs761237288 Chr.17: 37704902-37704904 on Build GRCh38
rs911202452 Chr.17: 37705039-37705039 on Build GRCh38
rs745986578 Chr.17: 37704919-37704919 on Build GRCh38
rs778125017 Chr.17: 37704902-37704902 on Build GRCh38
rs774350273 Chr.17: 37704956-37704956 on Build GRCh38
rs535041745 Chr.17: 37704911-37704911 on Build GRCh38
rs747280469 Chr.17: 37704909-37704909 on Build GRCh38
rs1051029550 Chr.17: 37704976-37704976 on Build GRCh38
rs772829938 Chr.17: 37704963-37704963 on Build GRCh38
rs2107133 Chr.17: 37704890-37704890 on Build GRCh38
rs764748514 Chr.17: 37705069-37705070 on Build GRCh38
rs199849203 Chr.17: 37705067-37705067 on Build GRCh38
rs753229331 Chr.17: 37704996-37704996 on Build GRCh38
rs757508365 Chr.17: 37705036-37705036 on Build GRCh38
rs765654692 Chr.17: 37705013-37705013 on Build GRCh38
rs369382627 Chr.17: 37705088-37705088 on Build GRCh38
rs537197692 Chr.17: 37705047-37705047 on Build GRCh38
rs753301333 Chr.17: 37705016-37705016 on Build GRCh38
rs898584035 Chr.17: 37705087-37705087 on Build GRCh38
rs760279499 Chr.17: 37704978-37704978 on Build GRCh38
rs747110790 Chr.17: 37705049-37705049 on Build GRCh38
rs141373830 Chr.17: 37704959-37704959 on Build GRCh38
rs751144953 Chr.17: 37704880-37704880 on Build GRCh38
rs879006831 Chr.17: 37705005-37705005 on Build GRCh38
rs772027416 Chr.17: 37705082-37705082 on Build GRCh38
rs754520966 Chr.17: 37704998-37704998 on Build GRCh38
rs111982126 Chr.17: 37705029-37705029 on Build GRCh38
rs747412954 Chr.17: 37705030-37705030 on Build GRCh38
rs74881740 Chr.17: 37704997-37704997 on Build GRCh38
rs768424318 Chr.17: 37704926-37704926 on Build GRCh38
rs193922482 Chr.17: 37704931-37704931 on Build GRCh38

View Full Product Details