Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241995998-241996271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP1LC3C
Entrez Gene ID
440738
Gene Name
microtubule associated protein 1 light chain 3 gamma
Gene Aliases
ATG8J, LC3C
UniGene
Hs.534971
Gene Chromosome Location
Chr.1: 241995194-242001393 on Build GRCh38
Transcripts
NM_001004343.2 XM_005273139.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6125603 c.360C>A p.A120A Chr.1: 241996247-241996247 on Build GRCh38
3486205 c.351G>A p.M117I Chr.1: 241996256-241996256 on Build GRCh38
4492298 c.395C>A p.A132D Chr.1: 241996212-241996212 on Build GRCh38
232754 c.362C>T p.S121F Chr.1: 241996245-241996245 on Build GRCh38
7090664 c.396C>A p.A132A Chr.1: 241996211-241996211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757525909 Chr.1: 241996122-241996122 on Build GRCh38
rs750797015 Chr.1: 241996193-241996193 on Build GRCh38
rs143833562 Chr.1: 241996168-241996168 on Build GRCh38
rs1020111743 Chr.1: 241996085-241996085 on Build GRCh38
rs889767872 Chr.1: 241996053-241996053 on Build GRCh38
rs562859017 Chr.1: 241996147-241996147 on Build GRCh38
rs575144045 Chr.1: 241996027-241996027 on Build GRCh38
rs763629571 Chr.1: 241996163-241996163 on Build GRCh38
rs191718681 Chr.1: 241996097-241996097 on Build GRCh38
rs959965374 Chr.1: 241996158-241996158 on Build GRCh38
rs779343268 Chr.1: 241996126-241996126 on Build GRCh38
rs1049737898 Chr.1: 241996052-241996052 on Build GRCh38
rs770562679 Chr.1: 241996152-241996152 on Build GRCh38
rs931537137 Chr.1: 241996028-241996028 on Build GRCh38
rs760152788 Chr.1: 241996161-241996161 on Build GRCh38
rs907620922 Chr.1: 241996186-241996186 on Build GRCh38
rs147920150 Chr.1: 241996250-241996250 on Build GRCh38
rs753535259 Chr.1: 241996116-241996116 on Build GRCh38
rs376488053 Chr.1: 241996246-241996246 on Build GRCh38
rs775442811 Chr.1: 241996159-241996159 on Build GRCh38
rs976622899 Chr.1: 241996108-241996108 on Build GRCh38
rs541125593 Chr.1: 241996063-241996063 on Build GRCh38
rs764859658 Chr.1: 241996178-241996178 on Build GRCh38
rs989017739 Chr.1: 241996047-241996047 on Build GRCh38
rs548700699 Chr.1: 241996129-241996129 on Build GRCh38
rs200884969 Chr.1: 241996154-241996154 on Build GRCh38
rs748433573 Chr.1: 241996148-241996148 on Build GRCh38
rs968797461 Chr.1: 241996059-241996059 on Build GRCh38
rs766971560 Chr.1: 241996212-241996212 on Build GRCh38
rs542437229 Chr.1: 241996079-241996079 on Build GRCh38
rs781425891 Chr.1: 241996247-241996247 on Build GRCh38
rs878968001 Chr.1: 241996111-241996111 on Build GRCh38
rs1010500201 Chr.1: 241996240-241996240 on Build GRCh38
rs540070974 Chr.1: 241996086-241996086 on Build GRCh38
rs200217615 Chr.1: 241996130-241996130 on Build GRCh38
rs202097500 Chr.1: 241996142-241996142 on Build GRCh38
rs28605139 Chr.1: 241996155-241996155 on Build GRCh38
rs747055229 Chr.1: 241996133-241996133 on Build GRCh38
rs768907198 Chr.1: 241996141-241996141 on Build GRCh38
rs111755040 Chr.1: 241996195-241996195 on Build GRCh38
rs537701934 Chr.1: 241996058-241996058 on Build GRCh38
rs752135369 Chr.1: 241996238-241996238 on Build GRCh38
rs56222938 Chr.1: 241996196-241996196 on Build GRCh38
rs147274936 Chr.1: 241996075-241996075 on Build GRCh38
rs1010491386 Chr.1: 241996120-241996120 on Build GRCh38
rs867743099 Chr.1: 241996249-241996249 on Build GRCh38

View Full Product Details