Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 8730792-8731019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIDINS220
Entrez Gene ID
57498
Gene Name
kinase D-interacting substrate 220kDa
Gene Aliases
ARMS
UniGene
Hs.9873
Gene Chromosome Location
Chr.2: 8728857-8837625 on Build GRCh38
Transcripts
NM_020738.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3583894 c.5046C>T p.T1682T Chr.2: 8730990-8730990 on Build GRCh38
6858714 c.5243A>T p.D1748V Chr.2: 8730793-8730793 on Build GRCh38
1752709 c.5128C>G p.Q1710E Chr.2: 8730908-8730908 on Build GRCh38
4398277 c.5038C>T p.P1680S Chr.2: 8730998-8730998 on Build GRCh38
722940 c.5031C>T p.N1677N Chr.2: 8731005-8731005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764865735 Chr.2: 8730930-8730930 on Build GRCh38
rs761325162 Chr.2: 8730816-8730816 on Build GRCh38
rs753065939 Chr.2: 8730965-8730965 on Build GRCh38
rs752145550 Chr.2: 8730949-8730949 on Build GRCh38
rs367740187 Chr.2: 8730989-8730989 on Build GRCh38
rs368542023 Chr.2: 8730885-8730885 on Build GRCh38
rs755413482 Chr.2: 8730830-8730830 on Build GRCh38
rs199639014 Chr.2: 8730996-8730996 on Build GRCh38
rs755379701 Chr.2: 8730952-8730952 on Build GRCh38
rs749481172 Chr.2: 8730971-8730971 on Build GRCh38
rs565122779 Chr.2: 8730845-8730845 on Build GRCh38
rs769837969 Chr.2: 8730839-8730839 on Build GRCh38
rs757356860 Chr.2: 8730974-8730974 on Build GRCh38
rs750086954 Chr.2: 8730939-8730939 on Build GRCh38
rs375496239 Chr.2: 8730829-8730829 on Build GRCh38
rs78365141 Chr.2: 8730968-8730968 on Build GRCh38
rs545714450 Chr.2: 8730840-8730840 on Build GRCh38
rs748413222 Chr.2: 8730836-8730836 on Build GRCh38
rs780769477 Chr.2: 8730826-8730826 on Build GRCh38
rs764819132 Chr.2: 8730817-8730817 on Build GRCh38
rs781621604 Chr.2: 8730960-8730960 on Build GRCh38
rs978044577 Chr.2: 8730915-8730915 on Build GRCh38
rs200371623 Chr.2: 8730831-8730831 on Build GRCh38
rs747472903 Chr.2: 8730852-8730852 on Build GRCh38
rs775445268 Chr.2: 8730868-8730868 on Build GRCh38
rs1010761756 Chr.2: 8730877-8730877 on Build GRCh38
rs201465267 Chr.2: 8730862-8730862 on Build GRCh38
rs76182610 Chr.2: 8730966-8730966 on Build GRCh38
rs762604646 Chr.2: 8730940-8730940 on Build GRCh38
rs749423918 Chr.2: 8730855-8730855 on Build GRCh38
rs768517761 Chr.2: 8730893-8730893 on Build GRCh38
rs372797696 Chr.2: 8730899-8730899 on Build GRCh38
rs749886554 Chr.2: 8730821-8730821 on Build GRCh38
rs527727658 Chr.2: 8730976-8730976 on Build GRCh38
rs761533158 Chr.2: 8730913-8730913 on Build GRCh38
rs1034611622 Chr.2: 8730936-8730936 on Build GRCh38
rs910289688 Chr.2: 8730953-8730953 on Build GRCh38
rs78490034 Chr.2: 8730967-8730967 on Build GRCh38
rs770976675 Chr.2: 8730858-8730858 on Build GRCh38
rs924892876 Chr.2: 8730928-8730928 on Build GRCh38
rs373610139 Chr.2: 8730944-8730944 on Build GRCh38
rs757977270 Chr.2: 8730822-8730822 on Build GRCh38
rs902303452 Chr.2: 8730901-8730901 on Build GRCh38
rs778914249 Chr.2: 8730981-8730981 on Build GRCh38
rs781374170 Chr.2: 8730835-8730835 on Build GRCh38

View Full Product Details