Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108606188-108606417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102B
Entrez Gene ID
284611
Gene Name
family with sequence similarity 102 member B
Gene Aliases
SYM-3B
UniGene
Hs.200230
Gene Chromosome Location
Chr.1: 108560349-108639327 on Build GRCh38
Transcripts
NM_001010883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936202 c.176G>C p.G59A Chr.1: 108606233-108606233 on Build GRCh38
4936201 c.269G>C p.G90A Chr.1: 108606233-108606233 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557160620 Chr.1: 108606228-108606228 on Build GRCh38
rs915673827 Chr.1: 108606360-108606360 on Build GRCh38
rs992512843 Chr.1: 108606354-108606354 on Build GRCh38
rs759774431 Chr.1: 108606274-108606274 on Build GRCh38
rs922020083 Chr.1: 108606279-108606279 on Build GRCh38
rs762864380 Chr.1: 108606226-108606226 on Build GRCh38
rs185912847 Chr.1: 108606303-108606303 on Build GRCh38
rs200751506 Chr.1: 108606294-108606294 on Build GRCh38
rs752820988 Chr.1: 108606296-108606296 on Build GRCh38
rs933307395 Chr.1: 108606286-108606286 on Build GRCh38
rs959874025 Chr.1: 108606345-108606345 on Build GRCh38
rs972045128 Chr.1: 108606300-108606300 on Build GRCh38
rs761671244 Chr.1: 108606249-108606249 on Build GRCh38
rs974816703 Chr.1: 108606262-108606262 on Build GRCh38
rs369875360 Chr.1: 108606272-108606272 on Build GRCh38
rs947886599 Chr.1: 108606384-108606384 on Build GRCh38
rs963919144 Chr.1: 108606258-108606258 on Build GRCh38
rs555063765 Chr.1: 108606302-108606302 on Build GRCh38
rs780669738 Chr.1: 108606371-108606371 on Build GRCh38
rs191761505 Chr.1: 108606366-108606366 on Build GRCh38
rs935238049 Chr.1: 108606336-108606336 on Build GRCh38
rs575713964 Chr.1: 108606263-108606263 on Build GRCh38
rs540935820 Chr.1: 108606356-108606356 on Build GRCh38
rs758559899 Chr.1: 108606248-108606248 on Build GRCh38

View Full Product Details