Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26692875-26693142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK3
Entrez Gene ID
3777
Gene Name
potassium two pore domain channel subfamily K member 3
Gene Aliases
K2p3.1, OAT1, PPH4, TASK, TASK-1, TBAK1
UniGene
Hs.24040
Gene Chromosome Location
Chr.2: 26692713-26733420 on Build GRCh38
Transcripts
NM_002246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4694492 c.177C>T p.Y59Y Chr.2: 26693052-26693052 on Build GRCh38
1255370 c.191G>A p.R64H Chr.2: 26693066-26693066 on Build GRCh38
1211576 c.110A>T p.E37V Chr.2: 26692985-26692985 on Build GRCh38
7373149 c.108C>G p.I36M Chr.2: 26692983-26692983 on Build GRCh38
6387459 c.240C>A p.F80L Chr.2: 26693115-26693115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761090652 Chr.2: 26693027-26693027 on Build GRCh38
rs977915621 Chr.2: 26693082-26693082 on Build GRCh38
rs144204029 Chr.2: 26693089-26693089 on Build GRCh38
rs1022064324 Chr.2: 26692995-26692995 on Build GRCh38
rs768129051 Chr.2: 26693008-26693008 on Build GRCh38
rs776045749 Chr.2: 26693022-26693022 on Build GRCh38
rs1010960867 Chr.2: 26692983-26692983 on Build GRCh38
rs528661453 Chr.2: 26692974-26692974 on Build GRCh38
rs770047414 Chr.2: 26692968-26692968 on Build GRCh38
rs535800063 Chr.2: 26692899-26692899 on Build GRCh38
rs746843850 Chr.2: 26693095-26693095 on Build GRCh38
rs550535933 Chr.2: 26692994-26692994 on Build GRCh38
rs568555516 Chr.2: 26693111-26693111 on Build GRCh38
rs753384871 Chr.2: 26693047-26693047 on Build GRCh38
rs773271941 Chr.2: 26692969-26692969 on Build GRCh38
rs749944072 Chr.2: 26693078-26693078 on Build GRCh38
rs1025063325 Chr.2: 26693077-26693077 on Build GRCh38
rs756194336 Chr.2: 26693093-26693094 on Build GRCh38
rs376610850 Chr.2: 26692978-26692978 on Build GRCh38
rs895163321 Chr.2: 26692930-26692930 on Build GRCh38
rs763871946 Chr.2: 26693046-26693046 on Build GRCh38
rs764584089 Chr.2: 26693070-26693070 on Build GRCh38
rs779940782 Chr.2: 26693090-26693090 on Build GRCh38
rs746788944 Chr.2: 26692959-26692959 on Build GRCh38
rs758193200 Chr.2: 26692912-26692912 on Build GRCh38
rs779891608 Chr.2: 26692915-26692915 on Build GRCh38

View Full Product Details