Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1212155-1212380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBP1
Entrez Gene ID
1487
Gene Name
C-terminal binding protein 1
Gene Aliases
BARS
UniGene
Hs.208597
Gene Chromosome Location
Chr.4: 1211440-1250329 on Build GRCh38
Transcripts
NM_001012614.1 NM_001328.2 XM_005272261.3 XM_005272263.4 XM_011513395.2 XM_011513396.2 XM_011513397.2 XM_017007762.1 XM_017007763.1 XM_017007764.1 XM_017007765.1 XM_017007766.1 XM_017007767.1 NR_104331.1

Gene Symbol
CTBP1-AS
Entrez Gene ID
285463
Gene Name
CTBP1 antisense RNA
Gene Aliases
PCAT10
UniGene
Hs.707614
Gene Chromosome Location
Chr.4: 1210120-1218591 on Build GRCh38
Transcripts
NM_001012614.1 NM_001328.2 XM_005272261.3 XM_005272263.4 XM_011513395.2 XM_011513396.2 XM_011513397.2 XM_017007762.1 XM_017007763.1 XM_017007764.1 XM_017007765.1 XM_017007766.1 XM_017007767.1 NR_104331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6426308 c.1277C>A p.T426N Chr.4: 1212286-1212286 on Build GRCh38
7241499 c.1248C>A p.H416Q Chr.4: 1212315-1212315 on Build GRCh38
3409030 c.1287C>G p.P429P Chr.4: 1212276-1212276 on Build GRCh38
6432206 c.1280T>C p.V427A Chr.4: 1212283-1212283 on Build GRCh38
4936535 c.1200C>T p.I400I Chr.4: 1212363-1212363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377619487 Chr.4: 1212316-1212316 on Build GRCh38
rs865862007 Chr.4: 1212214-1212214 on Build GRCh38
rs757056389 Chr.4: 1212258-1212258 on Build GRCh38
rs758074443 Chr.4: 1212353-1212353 on Build GRCh38
rs201436512 Chr.4: 1212323-1212323 on Build GRCh38
rs758690207 Chr.4: 1212336-1212336 on Build GRCh38
rs771269618 Chr.4: 1212295-1212295 on Build GRCh38
rs371763271 Chr.4: 1212213-1212213 on Build GRCh38
rs551713236 Chr.4: 1212189-1212189 on Build GRCh38
rs137891531 Chr.4: 1212339-1212339 on Build GRCh38
rs749654333 Chr.4: 1212288-1212288 on Build GRCh38
rs552180452 Chr.4: 1212235-1212235 on Build GRCh38
rs968456186 Chr.4: 1212236-1212236 on Build GRCh38
rs770287962 Chr.4: 1212285-1212285 on Build GRCh38
rs1019998811 Chr.4: 1212228-1212228 on Build GRCh38
rs1005969919 Chr.4: 1212196-1212196 on Build GRCh38
rs763759376 Chr.4: 1212253-1212253 on Build GRCh38
rs145193058 Chr.4: 1212257-1212257 on Build GRCh38
rs369378786 Chr.4: 1212276-1212276 on Build GRCh38
rs750024621 Chr.4: 1212342-1212342 on Build GRCh38
rs748609943 Chr.4: 1212284-1212284 on Build GRCh38
rs199614101 Chr.4: 1212301-1212301 on Build GRCh38
rs868723811 Chr.4: 1212275-1212275 on Build GRCh38
rs1047212435 Chr.4: 1212179-1212179 on Build GRCh38
rs375089554 Chr.4: 1212217-1212217 on Build GRCh38
rs764108337 Chr.4: 1212327-1212327 on Build GRCh38
rs369652853 Chr.4: 1212324-1212324 on Build GRCh38
rs115317882 Chr.4: 1212187-1212187 on Build GRCh38
rs757167464 Chr.4: 1212354-1212354 on Build GRCh38
rs775743153 Chr.4: 1212321-1212321 on Build GRCh38
rs369644553 Chr.4: 1212312-1212312 on Build GRCh38
rs760712247 Chr.4: 1212245-1212245 on Build GRCh38
rs752660628 Chr.4: 1212237-1212237 on Build GRCh38
rs368867475 Chr.4: 1212204-1212204 on Build GRCh38
rs926950501 Chr.4: 1212261-1212261 on Build GRCh38
rs532821312 Chr.4: 1212270-1212270 on Build GRCh38
rs766641046 Chr.4: 1212208-1212208 on Build GRCh38
rs200540863 Chr.4: 1212271-1212271 on Build GRCh38
rs536898821 Chr.4: 1212328-1212328 on Build GRCh38
rs1045466 Chr.4: 1212203-1212203 on Build GRCh38
rs777637511 Chr.4: 1212352-1212353 on Build GRCh38
rs767750268 Chr.4: 1212224-1212224 on Build GRCh38
rs978852365 Chr.4: 1212246-1212246 on Build GRCh38
rs763015951 Chr.4: 1212206-1212206 on Build GRCh38
rs998478862 Chr.4: 1212207-1212207 on Build GRCh38
rs868716600 Chr.4: 1212313-1212313 on Build GRCh38

View Full Product Details