Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177523862-177524125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM193B
Entrez Gene ID
54540
Gene Name
family with sequence similarity 193 member B
Gene Aliases
IRIZIO
UniGene
Hs.484289
Gene Chromosome Location
Chr.5: 177519789-177554585 on Build GRCh38
Transcripts
NM_001190946.2 NR_024019.3 XM_006714875.2 XM_006714876.1 XM_006714878.1 XM_006714879.3 XM_006714880.1 XM_006714884.1 XM_006714885.1 XM_011534572.1 XM_011534573.1 XM_011534574.1 XM_011534579.2 XM_011534580.1 XM_011534581.1 XM_011534582.1 XM_011534583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7321028 c.1406A>G p.K469R Chr.5: 177523960-177523960 on Build GRCh38
7187404 c.2198delA p.D733fs*41 Chr.5: 177524032-177524032 on Build GRCh38
5547342 c.1344_1345delCC p.L449fs*11 Chr.5: 177524021-177524022 on Build GRCh38
7187405 c.1334delA p.D445fs*41 Chr.5: 177524032-177524032 on Build GRCh38
5547341 c.2208_2209delCC p.L737fs*11 Chr.5: 177524021-177524022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767898073 Chr.5: 177524045-177524045 on Build GRCh38
rs766553702 Chr.5: 177524022-177524022 on Build GRCh38
rs577933094 Chr.5: 177524000-177524000 on Build GRCh38
rs778120065 Chr.5: 177524061-177524061 on Build GRCh38
rs749977233 Chr.5: 177524073-177524073 on Build GRCh38
rs892357725 Chr.5: 177524079-177524079 on Build GRCh38
rs778483158 Chr.5: 177523969-177523969 on Build GRCh38
rs767415098 Chr.5: 177523950-177523950 on Build GRCh38
rs1052462970 Chr.5: 177524074-177524074 on Build GRCh38
rs748597117 Chr.5: 177523998-177523998 on Build GRCh38
rs777041440 Chr.5: 177523910-177523910 on Build GRCh38
rs1026933710 Chr.5: 177524003-177524003 on Build GRCh38
rs751843687 Chr.5: 177523941-177523941 on Build GRCh38
rs777784106 Chr.5: 177523959-177523959 on Build GRCh38
rs757245014 Chr.5: 177524067-177524067 on Build GRCh38
rs772013784 Chr.5: 177524030-177524030 on Build GRCh38
rs775178111 Chr.5: 177524025-177524025 on Build GRCh38
rs773801582 Chr.5: 177524009-177524009 on Build GRCh38
rs755306731 Chr.5: 177523944-177523944 on Build GRCh38
rs753829988 Chr.5: 177524066-177524066 on Build GRCh38
rs1010805067 Chr.5: 177524098-177524098 on Build GRCh38
rs765815663 Chr.5: 177523927-177523927 on Build GRCh38
rs762882741 Chr.5: 177524014-177524014 on Build GRCh38
rs756799708 Chr.5: 177523967-177523967 on Build GRCh38
rs910088657 Chr.5: 177523887-177523887 on Build GRCh38
rs534451020 Chr.5: 177523914-177523914 on Build GRCh38
rs776951400 Chr.5: 177523991-177523991 on Build GRCh38
rs752576884 Chr.5: 177523952-177523952 on Build GRCh38
rs763868248 Chr.5: 177524059-177524059 on Build GRCh38
rs759799042 Chr.5: 177524030-177524030 on Build GRCh38
rs942762094 Chr.5: 177523899-177523899 on Build GRCh38
rs758025385 Chr.5: 177524082-177524082 on Build GRCh38
rs375666731 Chr.5: 177524055-177524055 on Build GRCh38
rs773144347 Chr.5: 177523928-177523928 on Build GRCh38
rs762545916 Chr.5: 177524102-177524102 on Build GRCh38
rs769026425 Chr.5: 177523907-177523907 on Build GRCh38
rs763116742 Chr.5: 177523936-177523936 on Build GRCh38
rs752383224 Chr.5: 177524047-177524047 on Build GRCh38
rs755980984 Chr.5: 177523956-177523956 on Build GRCh38
rs745509573 Chr.5: 177523978-177523978 on Build GRCh38
rs368416273 Chr.5: 177523938-177523938 on Build GRCh38
rs372183962 Chr.5: 177524028-177524028 on Build GRCh38
rs553079502 Chr.5: 177523962-177523962 on Build GRCh38
rs771782220 Chr.5: 177523979-177523979 on Build GRCh38

View Full Product Details