Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159808836-159809081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL6
Entrez Gene ID
343413
Gene Name
Fc receptor like 6
Gene Aliases
FcRH6
UniGene
Hs.196955
Gene Chromosome Location
Chr.1: 159800506-159816257 on Build GRCh38
Transcripts
NM_001004310.2 NM_001284217.1 XM_005245128.3 XM_005245129.3 XM_005245131.3 XM_006711292.3 XM_011509480.1 XM_011509481.2 XM_017001176.1 XM_017001177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368091 c.367C>T p.R123* Chr.1: 159809008-159809008 on Build GRCh38
2084711 c.412C>T p.P138S Chr.1: 159809053-159809053 on Build GRCh38
7323097 c.342G>A p.L114L Chr.1: 159808983-159808983 on Build GRCh38
1284952 c.413C>T p.P138L Chr.1: 159809054-159809054 on Build GRCh38
234710 c.435C>T p.L145L Chr.1: 159809076-159809076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138094040 Chr.1: 159809058-159809058 on Build GRCh38
rs753480080 Chr.1: 159808978-159808978 on Build GRCh38
rs771218895 Chr.1: 159809012-159809012 on Build GRCh38
rs764227693 Chr.1: 159809055-159809055 on Build GRCh38
rs770029254 Chr.1: 159808910-159808910 on Build GRCh38
rs370839083 Chr.1: 159808999-159808999 on Build GRCh38
rs143993814 Chr.1: 159809009-159809009 on Build GRCh38
rs376765049 Chr.1: 159808926-159808926 on Build GRCh38
rs1028205935 Chr.1: 159808860-159808860 on Build GRCh38
rs755344786 Chr.1: 159808990-159808990 on Build GRCh38
rs879840101 Chr.1: 159808868-159808868 on Build GRCh38
rs199834744 Chr.1: 159808974-159808974 on Build GRCh38
rs60865130 Chr.1: 159808968-159808968 on Build GRCh38
rs772069683 Chr.1: 159808962-159808962 on Build GRCh38
rs774199069 Chr.1: 159808939-159808939 on Build GRCh38
rs756692036 Chr.1: 159809054-159809054 on Build GRCh38
rs775406232 Chr.1: 159809027-159809027 on Build GRCh38
rs540301050 Chr.1: 159808908-159808908 on Build GRCh38
rs753196366 Chr.1: 159809053-159809053 on Build GRCh38
rs750826370 Chr.1: 159808862-159808862 on Build GRCh38
rs147235429 Chr.1: 159808980-159808980 on Build GRCh38
rs766589252 Chr.1: 159808981-159808981 on Build GRCh38
rs759936682 Chr.1: 159809051-159809051 on Build GRCh38
rs998437489 Chr.1: 159808859-159808859 on Build GRCh38
rs143079884 Chr.1: 159808988-159808988 on Build GRCh38
rs569208608 Chr.1: 159809052-159809052 on Build GRCh38
rs980680828 Chr.1: 159808931-159808931 on Build GRCh38
rs149327737 Chr.1: 159808964-159808964 on Build GRCh38
rs972814498 Chr.1: 159808878-159808878 on Build GRCh38
rs369915512 Chr.1: 159808929-159808929 on Build GRCh38
rs754250753 Chr.1: 159808945-159808945 on Build GRCh38
rs553092318 Chr.1: 159809051-159809052 on Build GRCh38
rs1042112147 Chr.1: 159809005-159809005 on Build GRCh38
rs572966856 Chr.1: 159808904-159808904 on Build GRCh38
rs772776972 Chr.1: 159809047-159809047 on Build GRCh38
rs781699582 Chr.1: 159808991-159808991 on Build GRCh38
rs779165798 Chr.1: 159809060-159809060 on Build GRCh38
rs73030817 Chr.1: 159808872-159808872 on Build GRCh38
rs745655616 Chr.1: 159809018-159809018 on Build GRCh38
rs753318478 Chr.1: 159809002-159809002 on Build GRCh38
rs778524052 Chr.1: 159809002-159809014 on Build GRCh38
rs771803248 Chr.1: 159809025-159809025 on Build GRCh38
rs151204274 Chr.1: 159809028-159809028 on Build GRCh38
rs945044818 Chr.1: 159809004-159809004 on Build GRCh38
rs145922130 Chr.1: 159809008-159809008 on Build GRCh38
rs961427412 Chr.1: 159808877-159808877 on Build GRCh38
rs12042065 Chr.1: 159808909-159808909 on Build GRCh38
rs764917537 Chr.1: 159809040-159809040 on Build GRCh38
rs778524602 Chr.1: 159809016-159809016 on Build GRCh38
rs140295172 Chr.1: 159809032-159809032 on Build GRCh38
rs1016923045 Chr.1: 159808876-159808876 on Build GRCh38
rs778136949 Chr.1: 159808914-159808914 on Build GRCh38
rs776125711 Chr.1: 159809030-159809030 on Build GRCh38

View Full Product Details