Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172448036-172448267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHSR
Entrez Gene ID
2693
Gene Name
growth hormone secretagogue receptor
Gene Aliases
GHDP
UniGene
Hs.130212
Gene Chromosome Location
Chr.3: 172443291-172448456 on Build GRCh38
Transcripts
NM_004122.2 NM_198407.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4905386 c.271C>T p.L91F Chr.3: 172448143-172448143 on Build GRCh38
7148299 c.265G>A p.D89N Chr.3: 172448149-172448149 on Build GRCh38
95917 c.270G>T p.L90L Chr.3: 172448144-172448144 on Build GRCh38
6545045 c.227C>A p.T76N Chr.3: 172448187-172448187 on Build GRCh38
5571565 c.151G>C p.V51L Chr.3: 172448263-172448263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745521758 Chr.3: 172448157-172448157 on Build GRCh38
rs200051419 Chr.3: 172448197-172448197 on Build GRCh38
rs202000049 Chr.3: 172448183-172448183 on Build GRCh38
rs762900969 Chr.3: 172448063-172448063 on Build GRCh38
rs2232167 Chr.3: 172448060-172448060 on Build GRCh38
rs896390704 Chr.3: 172448195-172448195 on Build GRCh38
rs779668138 Chr.3: 172448178-172448178 on Build GRCh38
rs984015452 Chr.3: 172448222-172448222 on Build GRCh38
rs766495914 Chr.3: 172448089-172448089 on Build GRCh38
rs963350574 Chr.3: 172448095-172448095 on Build GRCh38
rs912089357 Chr.3: 172448231-172448231 on Build GRCh38
rs778689494 Chr.3: 172448240-172448240 on Build GRCh38
rs753686167 Chr.3: 172448117-172448117 on Build GRCh38
rs202240311 Chr.3: 172448250-172448250 on Build GRCh38
rs766366395 Chr.3: 172448224-172448224 on Build GRCh38
rs750026836 Chr.3: 172448242-172448242 on Build GRCh38
rs995230051 Chr.3: 172448207-172448207 on Build GRCh38
rs753808890 Chr.3: 172448234-172448234 on Build GRCh38
rs201241515 Chr.3: 172448059-172448059 on Build GRCh38
rs561879798 Chr.3: 172448057-172448057 on Build GRCh38
rs950877188 Chr.3: 172448211-172448211 on Build GRCh38
rs969873003 Chr.3: 172448159-172448159 on Build GRCh38
rs756144492 Chr.3: 172448093-172448093 on Build GRCh38
rs757070976 Chr.3: 172448136-172448136 on Build GRCh38
rs778233231 Chr.3: 172448080-172448080 on Build GRCh38
rs779397565 Chr.3: 172448246-172448246 on Build GRCh38
rs777112021 Chr.3: 172448185-172448185 on Build GRCh38
rs200019512 Chr.3: 172448221-172448221 on Build GRCh38
rs200806469 Chr.3: 172448123-172448123 on Build GRCh38
rs79053943 Chr.3: 172448143-172448143 on Build GRCh38
rs200978184 Chr.3: 172448204-172448204 on Build GRCh38
rs199541220 Chr.3: 172448162-172448162 on Build GRCh38
rs762219121 Chr.3: 172448186-172448186 on Build GRCh38
rs982343230 Chr.3: 172448195-172448202 on Build GRCh38
rs763973161 Chr.3: 172448232-172448232 on Build GRCh38
rs200820156 Chr.3: 172448182-172448182 on Build GRCh38
rs946722733 Chr.3: 172448248-172448248 on Build GRCh38
rs760510238 Chr.3: 172448230-172448230 on Build GRCh38
rs495225 Chr.3: 172448243-172448243 on Build GRCh38
rs766399905 Chr.3: 172448152-172448154 on Build GRCh38
rs201379528 Chr.3: 172448210-172448210 on Build GRCh38
rs751701685 Chr.3: 172448228-172448228 on Build GRCh38
rs1012194455 Chr.3: 172448196-172448196 on Build GRCh38
rs752565297 Chr.3: 172448091-172448091 on Build GRCh38
rs777514924 Chr.3: 172448108-172448108 on Build GRCh38
rs756983145 Chr.3: 172448238-172448238 on Build GRCh38
rs374183783 Chr.3: 172448181-172448181 on Build GRCh38
rs966256034 Chr.3: 172448239-172448239 on Build GRCh38

View Full Product Details