Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87359267-87359498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
UniGene
Hs.125039
Gene Chromosome Location
Chr.7: 87345635-87399796 on Build GRCh38
Transcripts
NM_001143935.1 NM_001243745.1 NM_021151.3 XM_011516337.2 XM_017012370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7362738 c.188G>T p.G63V Chr.7: 87359278-87359278 on Build GRCh38
3726822 c.181delG p.I62fs*25 Chr.7: 87359271-87359271 on Build GRCh38
4674770 c.233G>T p.R78I Chr.7: 87359323-87359323 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765147539 Chr.7: 87359291-87359291 on Build GRCh38
rs756552422 Chr.7: 87359391-87359391 on Build GRCh38
rs1036305650 Chr.7: 87359335-87359335 on Build GRCh38
rs187779783 Chr.7: 87359347-87359347 on Build GRCh38
rs1047011233 Chr.7: 87359414-87359414 on Build GRCh38
rs544748910 Chr.7: 87359344-87359344 on Build GRCh38
rs762517024 Chr.7: 87359300-87359300 on Build GRCh38
rs373518254 Chr.7: 87359310-87359310 on Build GRCh38
rs112310560 Chr.7: 87359327-87359327 on Build GRCh38
rs780652599 Chr.7: 87359348-87359348 on Build GRCh38
rs1029236117 Chr.7: 87359365-87359365 on Build GRCh38
rs941180759 Chr.7: 87359465-87359465 on Build GRCh38
rs369915446 Chr.7: 87359341-87359341 on Build GRCh38
rs749724100 Chr.7: 87359404-87359404 on Build GRCh38
rs977198516 Chr.7: 87359462-87359462 on Build GRCh38
rs997718879 Chr.7: 87359362-87359362 on Build GRCh38
rs943439856 Chr.7: 87359352-87359352 on Build GRCh38
rs778111192 Chr.7: 87359396-87359396 on Build GRCh38
rs11384079 Chr.7: 87359354-87359355 on Build GRCh38
rs967504146 Chr.7: 87359428-87359428 on Build GRCh38
rs976507979 Chr.7: 87359361-87359361 on Build GRCh38
rs957533383 Chr.7: 87359323-87359323 on Build GRCh38
rs771421090 Chr.7: 87359354-87359356 on Build GRCh38
rs746599200 Chr.7: 87359374-87359377 on Build GRCh38
rs750186723 Chr.7: 87359295-87359295 on Build GRCh38
rs1012121465 Chr.7: 87359380-87359380 on Build GRCh38
rs753222362 Chr.7: 87359385-87359385 on Build GRCh38
rs889384805 Chr.7: 87359372-87359372 on Build GRCh38

View Full Product Details