Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19296085-19296336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4C
Entrez Gene ID
55667
Gene Name
DENN domain containing 4C
Gene Aliases
C9orf55, C9orf55B, RAB10GEF, bA513M16.3
UniGene
Hs.249591
Gene Chromosome Location
Chr.9: 19230765-19374268 on Build GRCh38
Transcripts
NM_017925.6 NR_073201.3 XM_006716804.3 XM_006716805.3 XM_011517959.2 XM_017014917.1 XM_017014918.1 XM_017014919.1 XM_017014920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6478434 c.191G>A p.R64K Chr.9: 19296105-19296105 on Build GRCh38
6190325 c.258delT p.F86fs*55 Chr.9: 19296172-19296172 on Build GRCh38
1461656 c.315T>C p.H105H Chr.9: 19296229-19296229 on Build GRCh38
6182932 c.242C>G p.S81* Chr.9: 19296156-19296156 on Build GRCh38
6376828 c.304T>C p.S102P Chr.9: 19296218-19296218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780041574 Chr.9: 19296117-19296117 on Build GRCh38
rs746475825 Chr.9: 19296286-19296286 on Build GRCh38
rs369054550 Chr.9: 19296195-19296195 on Build GRCh38
rs11788720 Chr.9: 19296277-19296277 on Build GRCh38
rs756424305 Chr.9: 19296237-19296237 on Build GRCh38
rs565735443 Chr.9: 19296212-19296212 on Build GRCh38
rs780381139 Chr.9: 19296241-19296241 on Build GRCh38
rs749040137 Chr.9: 19296120-19296120 on Build GRCh38
rs368908150 Chr.9: 19296161-19296164 on Build GRCh38
rs375001394 Chr.9: 19296133-19296133 on Build GRCh38
rs917098589 Chr.9: 19296179-19296179 on Build GRCh38
rs527969829 Chr.9: 19296272-19296272 on Build GRCh38
rs770602827 Chr.9: 19296290-19296290 on Build GRCh38
rs771674849 Chr.9: 19296141-19296141 on Build GRCh38
rs201208731 Chr.9: 19296213-19296213 on Build GRCh38
rs996335504 Chr.9: 19296301-19296301 on Build GRCh38
rs772597156 Chr.9: 19296157-19296157 on Build GRCh38
rs753687119 Chr.9: 19296253-19296253 on Build GRCh38
rs139831582 Chr.9: 19296259-19296259 on Build GRCh38
rs950807719 Chr.9: 19296299-19296299 on Build GRCh38
rs1054131724 Chr.9: 19296177-19296177 on Build GRCh38
rs995306255 Chr.9: 19296287-19296287 on Build GRCh38
rs377535488 Chr.9: 19296307-19296307 on Build GRCh38
rs898914437 Chr.9: 19296219-19296219 on Build GRCh38
rs764514703 Chr.9: 19296200-19296200 on Build GRCh38
rs201528021 Chr.9: 19296269-19296269 on Build GRCh38
rs199516367 Chr.9: 19296110-19296110 on Build GRCh38
rs935702552 Chr.9: 19296167-19296167 on Build GRCh38
rs1057407144 Chr.9: 19296160-19296160 on Build GRCh38
rs747840367 Chr.9: 19296288-19296289 on Build GRCh38
rs768741867 Chr.9: 19296126-19296126 on Build GRCh38
rs773919701 Chr.9: 19296129-19296129 on Build GRCh38
rs747621061 Chr.9: 19296132-19296132 on Build GRCh38
rs1027666938 Chr.9: 19296289-19296289 on Build GRCh38
rs771876601 Chr.9: 19296275-19296275 on Build GRCh38
rs375928640 Chr.9: 19296224-19296224 on Build GRCh38
rs776200314 Chr.9: 19296296-19296296 on Build GRCh38
rs752046601 Chr.9: 19296202-19296202 on Build GRCh38
rs760363457 Chr.9: 19296186-19296186 on Build GRCh38
rs770551577 Chr.9: 19296194-19296194 on Build GRCh38
rs773939635 Chr.9: 19296156-19296157 on Build GRCh38
rs749667003 Chr.9: 19296298-19296298 on Build GRCh38

View Full Product Details