Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10274742-10274985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICAM1
Entrez Gene ID
3383
Gene Name
intercellular adhesion molecule 1
Gene Aliases
BB2, CD54, P3.58
UniGene
Hs.643447
Gene Chromosome Location
Chr.19: 10270841-10286615 on Build GRCh38
Transcripts
NM_000201.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4949434 c.256G>T p.E86* Chr.19: 10274953-10274953 on Build GRCh38
6017234 c.193C>T p.P65S Chr.19: 10274890-10274890 on Build GRCh38
4984994 c.167A>T p.K56M Chr.19: 10274864-10274864 on Build GRCh38
5647313 c.275G>A p.C92Y Chr.19: 10274972-10274972 on Build GRCh38
3528123 c.96C>T p.S32S Chr.19: 10274793-10274793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143731465 Chr.19: 10274910-10274910 on Build GRCh38
rs759316086 Chr.19: 10274767-10274767 on Build GRCh38
rs759672075 Chr.19: 10274860-10274860 on Build GRCh38
rs747111380 Chr.19: 10274815-10274815 on Build GRCh38
rs554091291 Chr.19: 10274924-10274924 on Build GRCh38
rs767201668 Chr.19: 10274776-10274776 on Build GRCh38
rs992809768 Chr.19: 10274844-10274844 on Build GRCh38
rs5491 Chr.19: 10274864-10274864 on Build GRCh38
rs111997581 Chr.19: 10274887-10274887 on Build GRCh38
rs773878586 Chr.19: 10274852-10274852 on Build GRCh38
rs200156450 Chr.19: 10274841-10274841 on Build GRCh38
rs200970471 Chr.19: 10274858-10274858 on Build GRCh38
rs371538999 Chr.19: 10274904-10274904 on Build GRCh38
rs750328429 Chr.19: 10274844-10274845 on Build GRCh38
rs771818150 Chr.19: 10274827-10274827 on Build GRCh38
rs370020996 Chr.19: 10274886-10274886 on Build GRCh38
rs774261335 Chr.19: 10274930-10274930 on Build GRCh38
rs151201212 Chr.19: 10274835-10274835 on Build GRCh38
rs760537870 Chr.19: 10274883-10274883 on Build GRCh38
rs778474887 Chr.19: 10274818-10274818 on Build GRCh38
rs755971015 Chr.19: 10274803-10274803 on Build GRCh38
rs746006779 Chr.19: 10274947-10274947 on Build GRCh38
rs41520346 Chr.19: 10274790-10274790 on Build GRCh38
rs374718798 Chr.19: 10274915-10274915 on Build GRCh38
rs763087198 Chr.19: 10274854-10274854 on Build GRCh38
rs1041158342 Chr.19: 10274800-10274800 on Build GRCh38
rs535089993 Chr.19: 10274838-10274838 on Build GRCh38
rs1015821647 Chr.19: 10274862-10274862 on Build GRCh38
rs745524523 Chr.19: 10274826-10274826 on Build GRCh38
rs756786973 Chr.19: 10274816-10274816 on Build GRCh38
rs372704720 Chr.19: 10274919-10274919 on Build GRCh38
rs778384721 Chr.19: 10274901-10274901 on Build GRCh38
rs548184831 Chr.19: 10274859-10274859 on Build GRCh38
rs146780348 Chr.19: 10274923-10274923 on Build GRCh38
rs763975382 Chr.19: 10274885-10274885 on Build GRCh38
rs781408937 Chr.19: 10274832-10274832 on Build GRCh38
rs369268780 Chr.19: 10274918-10274918 on Build GRCh38
rs1032707496 Chr.19: 10274945-10274945 on Build GRCh38
rs137977632 Chr.19: 10274812-10274812 on Build GRCh38
rs190348035 Chr.19: 10274928-10274928 on Build GRCh38

View Full Product Details