Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96489875-96490136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HABP4
Entrez Gene ID
22927
Gene Name
hyaluronan binding protein 4
Gene Aliases
IHABP-4, IHABP4, Ki-1/57, SERBP1L
UniGene
Hs.494567
Gene Chromosome Location
Chr.9: 96450155-96491336 on Build GRCh38
Transcripts
NM_014282.3 XM_005251812.3 XM_017014484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238651 c.1218G>T p.P406P Chr.9: 96490014-96490014 on Build GRCh38
1111618 c.1233G>A p.A411A Chr.9: 96490029-96490029 on Build GRCh38
5767611 c.1201C>A p.P401T Chr.9: 96489997-96489997 on Build GRCh38
1490243 c.1218G>A p.P406P Chr.9: 96490014-96490014 on Build GRCh38
4461941 c.1227C>T p.F409F Chr.9: 96490023-96490023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767661319 Chr.9: 96490058-96490058 on Build GRCh38
rs373559224 Chr.9: 96489971-96489971 on Build GRCh38
rs368414676 Chr.9: 96489930-96489930 on Build GRCh38
rs1041499097 Chr.9: 96489925-96489925 on Build GRCh38
rs778144301 Chr.9: 96489968-96489968 on Build GRCh38
rs770555199 Chr.9: 96489985-96489985 on Build GRCh38
rs912593053 Chr.9: 96490106-96490106 on Build GRCh38
rs376355262 Chr.9: 96490097-96490097 on Build GRCh38
rs536215022 Chr.9: 96489895-96489895 on Build GRCh38
rs12347119 Chr.9: 96490060-96490060 on Build GRCh38
rs755233832 Chr.9: 96489942-96489942 on Build GRCh38
rs761804436 Chr.9: 96490016-96490016 on Build GRCh38
rs376431061 Chr.9: 96490029-96490029 on Build GRCh38
rs766412891 Chr.9: 96490028-96490028 on Build GRCh38
rs893295823 Chr.9: 96489918-96489918 on Build GRCh38
rs745488619 Chr.9: 96489987-96489987 on Build GRCh38
rs756759174 Chr.9: 96490059-96490059 on Build GRCh38
rs757883164 Chr.9: 96490071-96490071 on Build GRCh38
rs776447402 Chr.9: 96490015-96490015 on Build GRCh38
rs372203280 Chr.9: 96489913-96489913 on Build GRCh38
rs527438879 Chr.9: 96490033-96490033 on Build GRCh38
rs201262154 Chr.9: 96489932-96489932 on Build GRCh38
rs368896005 Chr.9: 96489946-96489946 on Build GRCh38
rs529464025 Chr.9: 96490079-96490079 on Build GRCh38
rs199959448 Chr.9: 96490077-96490077 on Build GRCh38
rs963557212 Chr.9: 96490002-96490002 on Build GRCh38
rs919141065 Chr.9: 96489903-96489903 on Build GRCh38
rs190257474 Chr.9: 96489921-96489921 on Build GRCh38
rs779413318 Chr.9: 96490044-96490044 on Build GRCh38
rs778261333 Chr.9: 96490061-96490061 on Build GRCh38
rs374794660 Chr.9: 96489931-96489931 on Build GRCh38
rs766502963 Chr.9: 96489933-96489933 on Build GRCh38
rs534274476 Chr.9: 96490050-96490050 on Build GRCh38
rs751774503 Chr.9: 96489941-96489941 on Build GRCh38
rs760418339 Chr.9: 96490001-96490001 on Build GRCh38
rs765729919 Chr.9: 96490062-96490063 on Build GRCh38
rs759551644 Chr.9: 96489965-96489972 on Build GRCh38
rs768573343 Chr.9: 96490014-96490014 on Build GRCh38
rs756600286 Chr.9: 96489950-96489950 on Build GRCh38
rs754394171 Chr.9: 96490062-96490062 on Build GRCh38
rs775306763 Chr.9: 96489991-96489991 on Build GRCh38
rs182885483 Chr.9: 96489986-96489986 on Build GRCh38
rs771902546 Chr.9: 96489989-96489989 on Build GRCh38

View Full Product Details