Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42786686-42786935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM216B
Entrez Gene ID
144809
Gene Name
family with sequence similarity 216 member B
Gene Aliases
C13orf30
UniGene
Hs.16970
Gene Chromosome Location
Chr.13: 42781550-42791549 on Build GRCh38
Transcripts
NM_001318932.1 NM_182508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4433642 c.108C>A p.N36K Chr.13: 42786771-42786771 on Build GRCh38
6553932 c.167G>T p.W56L Chr.13: 42786830-42786830 on Build GRCh38
4641044 c.100G>C p.A34P Chr.13: 42786763-42786763 on Build GRCh38
5376765 c.125A>T p.Y42F Chr.13: 42786788-42786788 on Build GRCh38
4927295 c.112G>A p.G38R Chr.13: 42786775-42786775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756007090 Chr.13: 42786848-42786848 on Build GRCh38
rs370728261 Chr.13: 42786832-42786832 on Build GRCh38
rs373916762 Chr.13: 42786897-42786897 on Build GRCh38
rs180825862 Chr.13: 42786888-42786888 on Build GRCh38
rs757825686 Chr.13: 42786876-42786876 on Build GRCh38
rs780992016 Chr.13: 42786766-42786766 on Build GRCh38
rs17063717 Chr.13: 42786898-42786898 on Build GRCh38
rs146856401 Chr.13: 42786827-42786827 on Build GRCh38
rs755173069 Chr.13: 42786779-42786779 on Build GRCh38
rs769325742 Chr.13: 42786817-42786817 on Build GRCh38
rs927332025 Chr.13: 42786837-42786837 on Build GRCh38
rs764748088 Chr.13: 42786750-42786750 on Build GRCh38
rs767246902 Chr.13: 42786835-42786835 on Build GRCh38
rs959029344 Chr.13: 42786847-42786847 on Build GRCh38
rs777148622 Chr.13: 42786804-42786804 on Build GRCh38
rs753015049 Chr.13: 42786853-42786853 on Build GRCh38
rs775246700 Chr.13: 42786821-42786821 on Build GRCh38
rs771383014 Chr.13: 42786798-42786798 on Build GRCh38
rs779487010 Chr.13: 42786829-42786829 on Build GRCh38
rs369392903 Chr.13: 42786736-42786736 on Build GRCh38
rs929910707 Chr.13: 42786731-42786731 on Build GRCh38
rs202087551 Chr.13: 42786866-42786866 on Build GRCh38
rs758361979 Chr.13: 42786861-42786861 on Build GRCh38
rs76544567 Chr.13: 42786904-42786904 on Build GRCh38
rs375560512 Chr.13: 42786908-42786908 on Build GRCh38
rs773921211 Chr.13: 42786721-42786721 on Build GRCh38
rs763909321 Chr.13: 42786759-42786759 on Build GRCh38
rs773243374 Chr.13: 42786909-42786909 on Build GRCh38
rs748997724 Chr.13: 42786901-42786901 on Build GRCh38
rs762593334 Chr.13: 42786753-42786753 on Build GRCh38
rs530475946 Chr.13: 42786844-42786844 on Build GRCh38
rs762414351 Chr.13: 42786745-42786745 on Build GRCh38
rs569905875 Chr.13: 42786871-42786871 on Build GRCh38
rs759266480 Chr.13: 42786746-42786746 on Build GRCh38
rs747541898 Chr.13: 42786795-42786795 on Build GRCh38
rs373692405 Chr.13: 42786760-42786760 on Build GRCh38
rs753997695 Chr.13: 42786777-42786777 on Build GRCh38
rs760002262 Chr.13: 42786806-42786806 on Build GRCh38
rs765348725 Chr.13: 42786850-42786850 on Build GRCh38
rs200110453 Chr.13: 42786784-42786784 on Build GRCh38
rs537911529 Chr.13: 42786903-42786903 on Build GRCh38
rs142430750 Chr.13: 42786786-42786786 on Build GRCh38
rs761088591 Chr.13: 42786724-42786724 on Build GRCh38
rs776373979 Chr.13: 42786745-42786745 on Build GRCh38
rs756993507 Chr.13: 42786764-42786764 on Build GRCh38
rs772235181 Chr.13: 42786785-42786785 on Build GRCh38
rs762345009 Chr.13: 42786824-42786824 on Build GRCh38
rs781781091 Chr.13: 42786885-42786885 on Build GRCh38
rs571342887 Chr.13: 42786782-42786782 on Build GRCh38

View Full Product Details