Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131182106-131182379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYRM7
Entrez Gene ID
90624
Gene Name
LYR motif containing 7
Gene Aliases
C5orf31, MC3DN8, MZM1L
UniGene
Hs.115467
Gene Chromosome Location
Chr.5: 131170914-131205426 on Build GRCh38
Transcripts
NM_001293735.1 NM_181705.3 NR_121658.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4466538 c.143C>T p.S48F Chr.5: 131182280-131182280 on Build GRCh38
1060518 c.155T>C p.I52T Chr.5: 131182292-131182292 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs79908895 Chr.5: 131182159-131182159 on Build GRCh38
rs909888327 Chr.5: 131182276-131182276 on Build GRCh38
rs762075975 Chr.5: 131182277-131182277 on Build GRCh38
rs750902702 Chr.5: 131182330-131182331 on Build GRCh38
rs777396016 Chr.5: 131182335-131182335 on Build GRCh38
rs573476169 Chr.5: 131182300-131182300 on Build GRCh38
rs138726230 Chr.5: 131182198-131182198 on Build GRCh38
rs756054444 Chr.5: 131182197-131182197 on Build GRCh38
rs762109952 Chr.5: 131182277-131182279 on Build GRCh38
rs373893622 Chr.5: 131182287-131182287 on Build GRCh38
rs753743067 Chr.5: 131182315-131182315 on Build GRCh38
rs905077983 Chr.5: 131182328-131182328 on Build GRCh38
rs767824191 Chr.5: 131182280-131182280 on Build GRCh38
rs748808798 Chr.5: 131182199-131182199 on Build GRCh38
rs375129014 Chr.5: 131182209-131182209 on Build GRCh38
rs754452398 Chr.5: 131182208-131182208 on Build GRCh38
rs778350015 Chr.5: 131182317-131182317 on Build GRCh38
rs541638565 Chr.5: 131182211-131182211 on Build GRCh38
rs762952204 Chr.5: 131182258-131182258 on Build GRCh38
rs752183737 Chr.5: 131182324-131182324 on Build GRCh38
rs535747849 Chr.5: 131182145-131182145 on Build GRCh38
rs774745145 Chr.5: 131182268-131182268 on Build GRCh38
rs558428692 Chr.5: 131182213-131182213 on Build GRCh38
rs771310136 Chr.5: 131182214-131182214 on Build GRCh38
rs750405795 Chr.5: 131182185-131182185 on Build GRCh38
rs762725490 Chr.5: 131182316-131182316 on Build GRCh38
rs767928754 Chr.5: 131182318-131182318 on Build GRCh38
rs368070144 Chr.5: 131182333-131182333 on Build GRCh38
rs200336982 Chr.5: 131182237-131182237 on Build GRCh38
rs764166987 Chr.5: 131182267-131182267 on Build GRCh38
rs542418348 Chr.5: 131182346-131182346 on Build GRCh38
rs758941942 Chr.5: 131182232-131182232 on Build GRCh38
rs766241507 Chr.5: 131182297-131182297 on Build GRCh38
rs767976577 Chr.5: 131182310-131182311 on Build GRCh38
rs750311709 Chr.5: 131182281-131182281 on Build GRCh38
rs199715117 Chr.5: 131182251-131182251 on Build GRCh38
rs746197873 Chr.5: 131182233-131182233 on Build GRCh38

View Full Product Details