Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39423242-39423467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCA5L
Entrez Gene ID
150082
Gene Name
LCA5L, lebercilin like
Gene Aliases
C21orf13
UniGene
Hs.517284
Gene Chromosome Location
Chr.21: 39405844-39445796 on Build GRCh38
Transcripts
NM_152505.3 XM_005260926.1 XM_006723967.1 XM_006723972.1 XM_006723973.1 XM_011529458.1 XM_011529459.1 XM_011529460.1 XM_011529461.1 XM_011529462.1 XM_011529463.2 XM_011529464.1 XM_011529465.1 XM_011529466.1 XM_011529467.1 XM_011529468.1 XM_011529469.2 XM_011529470.1 XM_011529471.1 XM_017028274.1 XM_017028275.1 XM_017028276.1 XM_017028277.1 XM_017028278.1 NM_001317744.1

Gene Symbol
WRB-SH3BGR
Entrez Gene ID
106865373
Gene Name
WRB-SH3BGR readthrough
Gene Aliases
-
UniGene
Hs.473847
Gene Chromosome Location
Chr.21: 39380244-39515506 on Build GRCh38
Transcripts
NM_152505.3 XM_005260926.1 XM_006723967.1 XM_006723972.1 XM_006723973.1 XM_011529458.1 XM_011529459.1 XM_011529460.1 XM_011529461.1 XM_011529462.1 XM_011529463.2 XM_011529464.1 XM_011529465.1 XM_011529466.1 XM_011529467.1 XM_011529468.1 XM_011529469.2 XM_011529470.1 XM_011529471.1 XM_017028274.1 XM_017028275.1 XM_017028276.1 XM_017028277.1 XM_017028278.1 NM_001317744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4611298 c.350_351insA p.H118fs*9 Chr.21: 39423462-39423463 on Build GRCh38
1751646 c.432C>T p.L144L Chr.21: 39423381-39423381 on Build GRCh38
461348 c.549G>C p.L183F Chr.21: 39423264-39423264 on Build GRCh38
234897 c.558A>C p.I186I Chr.21: 39423255-39423255 on Build GRCh38
7197049 c.441G>A p.R147R Chr.21: 39423372-39423372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751930333 Chr.21: 39423287-39423287 on Build GRCh38
rs749881986 Chr.21: 39423411-39423411 on Build GRCh38
rs947353048 Chr.21: 39423404-39423404 on Build GRCh38
rs763762859 Chr.21: 39423275-39423275 on Build GRCh38
rs572774636 Chr.21: 39423407-39423407 on Build GRCh38
rs141251948 Chr.21: 39423345-39423345 on Build GRCh38
rs146536316 Chr.21: 39423288-39423288 on Build GRCh38
rs138035610 Chr.21: 39423420-39423420 on Build GRCh38
rs1010372553 Chr.21: 39423282-39423282 on Build GRCh38
rs748496581 Chr.21: 39423314-39423314 on Build GRCh38
rs541698626 Chr.21: 39423418-39423418 on Build GRCh38
rs372477670 Chr.21: 39423281-39423281 on Build GRCh38
rs770244511 Chr.21: 39423317-39423317 on Build GRCh38
rs778163850 Chr.21: 39423329-39423329 on Build GRCh38
rs752179095 Chr.21: 39423294-39423294 on Build GRCh38
rs771872757 Chr.21: 39423372-39423372 on Build GRCh38
rs762805925 Chr.21: 39423419-39423419 on Build GRCh38
rs184105746 Chr.21: 39423338-39423338 on Build GRCh38
rs753208196 Chr.21: 39423430-39423430 on Build GRCh38
rs141252242 Chr.21: 39423388-39423388 on Build GRCh38
rs749466145 Chr.21: 39423331-39423331 on Build GRCh38
rs755430643 Chr.21: 39423302-39423302 on Build GRCh38
rs774279740 Chr.21: 39423339-39423339 on Build GRCh38
rs1034673261 Chr.21: 39423431-39423431 on Build GRCh38
rs150338806 Chr.21: 39423394-39423394 on Build GRCh38
rs200966361 Chr.21: 39423346-39423346 on Build GRCh38
rs747213855 Chr.21: 39423346-39423346 on Build GRCh38
rs188824462 Chr.21: 39423392-39423392 on Build GRCh38
rs760359609 Chr.21: 39423274-39423274 on Build GRCh38
rs138782916 Chr.21: 39423376-39423376 on Build GRCh38
rs144859622 Chr.21: 39423359-39423359 on Build GRCh38
rs781660376 Chr.21: 39423309-39423309 on Build GRCh38

View Full Product Details