Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61839347-61839572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM161A
Entrez Gene ID
84140
Gene Name
family with sequence similarity 161 member A
Gene Aliases
RP28
UniGene
Hs.440466
Gene Chromosome Location
Chr.2: 61800240-61856074 on Build GRCh38
Transcripts
NM_001201543.1 NM_032180.2 NR_037710.1 XM_017005072.1 XM_017005073.1 XM_017005074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658742 c.1550C>T p.T517M Chr.2: 61839454-61839454 on Build GRCh38
3910479 c.1540C>T p.P514S Chr.2: 61839464-61839464 on Build GRCh38
3054607 c.1460G>A p.R487H Chr.2: 61839544-61839544 on Build GRCh38
1306920 c.1548C>A p.P516P Chr.2: 61839456-61839456 on Build GRCh38
6658743 c.1550C>T p.T517M Chr.2: 61839454-61839454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754881620 Chr.2: 61839405-61839405 on Build GRCh38
rs765178656 Chr.2: 61839406-61839406 on Build GRCh38
rs769276393 Chr.2: 61839505-61839505 on Build GRCh38
rs961161022 Chr.2: 61839456-61839457 on Build GRCh38
rs200160715 Chr.2: 61839408-61839408 on Build GRCh38
rs1057332357 Chr.2: 61839384-61839384 on Build GRCh38
rs748311212 Chr.2: 61839518-61839518 on Build GRCh38
rs762767781 Chr.2: 61839529-61839529 on Build GRCh38
rs776174179 Chr.2: 61839426-61839426 on Build GRCh38
rs202193201 Chr.2: 61839437-61839437 on Build GRCh38
rs753702594 Chr.2: 61839391-61839391 on Build GRCh38
rs761440783 Chr.2: 61839540-61839540 on Build GRCh38
rs769560785 Chr.2: 61839523-61839523 on Build GRCh38
rs758465547 Chr.2: 61839473-61839474 on Build GRCh38
rs767414973 Chr.2: 61839503-61839503 on Build GRCh38
rs766501113 Chr.2: 61839475-61839479 on Build GRCh38
rs372121322 Chr.2: 61839425-61839425 on Build GRCh38
rs751120519 Chr.2: 61839461-61839461 on Build GRCh38
rs752639878 Chr.2: 61839450-61839450 on Build GRCh38
rs759263986 Chr.2: 61839521-61839521 on Build GRCh38
rs766078167 Chr.2: 61839388-61839388 on Build GRCh38
rs756552796 Chr.2: 61839470-61839470 on Build GRCh38
rs200331923 Chr.2: 61839431-61839431 on Build GRCh38
rs551187535 Chr.2: 61839531-61839531 on Build GRCh38
rs539389330 Chr.2: 61839462-61839462 on Build GRCh38
rs757001860 Chr.2: 61839419-61839419 on Build GRCh38
rs763507072 Chr.2: 61839454-61839454 on Build GRCh38
rs778830779 Chr.2: 61839511-61839511 on Build GRCh38
rs780651086 Chr.2: 61839499-61839499 on Build GRCh38
rs761940462 Chr.2: 61839434-61839434 on Build GRCh38
rs201998260 Chr.2: 61839520-61839520 on Build GRCh38
rs202003151 Chr.2: 61839389-61839390 on Build GRCh38
rs868604211 Chr.2: 61839526-61839526 on Build GRCh38
rs541823159 Chr.2: 61839490-61839490 on Build GRCh38
rs143622468 Chr.2: 61839453-61839453 on Build GRCh38
rs767768523 Chr.2: 61839436-61839436 on Build GRCh38
rs765389610 Chr.2: 61839410-61839413 on Build GRCh38
rs750983510 Chr.2: 61839538-61839538 on Build GRCh38
rs768632779 Chr.2: 61839428-61839428 on Build GRCh38
rs528505050 Chr.2: 61839417-61839417 on Build GRCh38
rs368458156 Chr.2: 61839410-61839410 on Build GRCh38
rs775421195 Chr.2: 61839527-61839527 on Build GRCh38
rs781661338 Chr.2: 61839420-61839420 on Build GRCh38
rs750449050 Chr.2: 61839457-61839469 on Build GRCh38
rs749749731 Chr.2: 61839503-61839503 on Build GRCh38

View Full Product Details