Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45888568-45888807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFBP1
Entrez Gene ID
3484
Gene Name
insulin like growth factor binding protein 1
Gene Aliases
AFBP, IBP1, IGF-BP25, PP12, hIGFBP-1
UniGene
Hs.642938
Gene Chromosome Location
Chr.7: 45888360-45893668 on Build GRCh38
Transcripts
NM_000596.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6892093 c.71C>A p.A24D Chr.7: 45888723-45888723 on Build GRCh38
1179375 c.41T>G p.L14R Chr.7: 45888693-45888693 on Build GRCh38
3832889 c.102C>T p.S34S Chr.7: 45888754-45888754 on Build GRCh38
6281821 c.141G>T p.S47S Chr.7: 45888793-45888793 on Build GRCh38
3881348 c.27C>T p.V9V Chr.7: 45888679-45888679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766860058 Chr.7: 45888736-45888736 on Build GRCh38
rs771841850 Chr.7: 45888697-45888697 on Build GRCh38
rs376937815 Chr.7: 45888768-45888768 on Build GRCh38
rs764291762 Chr.7: 45888626-45888626 on Build GRCh38
rs776759256 Chr.7: 45888717-45888717 on Build GRCh38
rs1025559126 Chr.7: 45888727-45888727 on Build GRCh38
rs755624791 Chr.7: 45888742-45888742 on Build GRCh38
rs772637342 Chr.7: 45888630-45888631 on Build GRCh38
rs769935884 Chr.7: 45888788-45888788 on Build GRCh38
rs762033595 Chr.7: 45888782-45888782 on Build GRCh38
rs750959927 Chr.7: 45888643-45888643 on Build GRCh38
rs148297000 Chr.7: 45888669-45888669 on Build GRCh38
rs555273668 Chr.7: 45888757-45888757 on Build GRCh38
rs371930012 Chr.7: 45888624-45888624 on Build GRCh38
rs745725926 Chr.7: 45888762-45888762 on Build GRCh38
rs761982277 Chr.7: 45888723-45888723 on Build GRCh38
rs369421475 Chr.7: 45888645-45888645 on Build GRCh38
rs748974940 Chr.7: 45888751-45888751 on Build GRCh38
rs754078165 Chr.7: 45888628-45888628 on Build GRCh38
rs777300659 Chr.7: 45888743-45888743 on Build GRCh38
rs111255251 Chr.7: 45888677-45888677 on Build GRCh38
rs773553957 Chr.7: 45888778-45888778 on Build GRCh38
rs138841724 Chr.7: 45888711-45888711 on Build GRCh38
rs758909946 Chr.7: 45888732-45888732 on Build GRCh38
rs777344533 Chr.7: 45888674-45888674 on Build GRCh38
rs765612314 Chr.7: 45888731-45888731 on Build GRCh38
rs147051230 Chr.7: 45888685-45888685 on Build GRCh38
rs780766478 Chr.7: 45888651-45888651 on Build GRCh38
rs745815847 Chr.7: 45888694-45888694 on Build GRCh38
rs570379585 Chr.7: 45888767-45888767 on Build GRCh38
rs373659181 Chr.7: 45888739-45888739 on Build GRCh38
rs774475002 Chr.7: 45888618-45888618 on Build GRCh38
rs775339662 Chr.7: 45888770-45888770 on Build GRCh38
rs369137225 Chr.7: 45888620-45888620 on Build GRCh38
rs776642572 Chr.7: 45888781-45888781 on Build GRCh38
rs377316369 Chr.7: 45888778-45888778 on Build GRCh38
rs760725872 Chr.7: 45888709-45888709 on Build GRCh38
rs1033685071 Chr.7: 45888707-45888707 on Build GRCh38
rs765555012 Chr.7: 45888638-45888638 on Build GRCh38
rs891666742 Chr.7: 45888639-45888639 on Build GRCh38
rs775494697 Chr.7: 45888704-45888704 on Build GRCh38
rs200116651 Chr.7: 45888625-45888625 on Build GRCh38
rs757668092 Chr.7: 45888634-45888634 on Build GRCh38
rs1013993759 Chr.7: 45888699-45888699 on Build GRCh38
rs746935293 Chr.7: 45888774-45888774 on Build GRCh38
rs770884467 Chr.7: 45888681-45888681 on Build GRCh38
rs747541587 Chr.7: 45888665-45888665 on Build GRCh38

View Full Product Details