Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169084862-169085135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MECOM
Entrez Gene ID
2122
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
UniGene
Hs.744090
Gene Chromosome Location
Chr.3: 169083499-169663781 on Build GRCh38
Transcripts
NM_001105077.3 NM_001105078.3 NM_001163999.1 NM_001164000.1 NM_001205194.1 NM_004991.3 NM_005241.3 XM_005247213.3 XM_005247214.3 XM_005247215.3 XM_005247219.2 XM_005247220.2 XM_005247221.2 XM_005247223.2 XM_005247224.3 XM_005247225.3 XM_005247226.3 XM_011512546.2 XM_017005874.1 XM_017005875.1 XM_017005876.1 XM_017005877.1 XM_017005878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
479689 c.3095G>A p.W1032* Chr.3: 169084970-169084970 on Build GRCh38
5829616 c.3657G>A p.A1219A Chr.3: 169084945-169084945 on Build GRCh38
1632968 c.3029C>T p.A1010V Chr.3: 169085036-169085036 on Build GRCh38
98030 c.3130G>T p.A1044S Chr.3: 169084935-169084935 on Build GRCh38
4856985 c.3290G>A p.W1097* Chr.3: 169084970-169084970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759485296 Chr.3: 169084994-169084994 on Build GRCh38
rs774280360 Chr.3: 169084902-169084902 on Build GRCh38
rs144104039 Chr.3: 169085028-169085028 on Build GRCh38
rs375898498 Chr.3: 169084896-169084896 on Build GRCh38
rs577871113 Chr.3: 169085104-169085104 on Build GRCh38
rs781425049 Chr.3: 169085079-169085079 on Build GRCh38
rs146810574 Chr.3: 169085029-169085029 on Build GRCh38
rs372906440 Chr.3: 169084914-169084914 on Build GRCh38
rs747301473 Chr.3: 169084959-169084959 on Build GRCh38
rs867621042 Chr.3: 169084938-169084938 on Build GRCh38
rs771547877 Chr.3: 169085006-169085006 on Build GRCh38
rs201825977 Chr.3: 169084949-169084949 on Build GRCh38
rs148539966 Chr.3: 169084993-169084993 on Build GRCh38
rs370609194 Chr.3: 169084908-169084908 on Build GRCh38
rs537452650 Chr.3: 169085013-169085013 on Build GRCh38
rs751652068 Chr.3: 169085076-169085076 on Build GRCh38
rs750937460 Chr.3: 169085049-169085049 on Build GRCh38
rs756269286 Chr.3: 169084920-169084920 on Build GRCh38
rs750090830 Chr.3: 169084945-169084945 on Build GRCh38
rs754263328 Chr.3: 169084918-169084918 on Build GRCh38
rs764070754 Chr.3: 169085016-169085019 on Build GRCh38
rs770720558 Chr.3: 169084985-169084985 on Build GRCh38
rs779316050 Chr.3: 169084946-169084946 on Build GRCh38
rs201640456 Chr.3: 169084915-169084915 on Build GRCh38
rs749087441 Chr.3: 169084975-169084975 on Build GRCh38
rs998597774 Chr.3: 169084933-169084933 on Build GRCh38
rs370701880 Chr.3: 169084955-169084955 on Build GRCh38
rs869220573 Chr.3: 169085079-169085079 on Build GRCh38
rs369748515 Chr.3: 169084912-169084912 on Build GRCh38
rs754463603 Chr.3: 169085088-169085088 on Build GRCh38
rs144184901 Chr.3: 169085038-169085038 on Build GRCh38
rs1005673341 Chr.3: 169084904-169084904 on Build GRCh38
rs758989032 Chr.3: 169084907-169084907 on Build GRCh38
rs750896996 Chr.3: 169084948-169084948 on Build GRCh38
rs375891441 Chr.3: 169085056-169085056 on Build GRCh38
rs748346216 Chr.3: 169085091-169085091 on Build GRCh38
rs979988402 Chr.3: 169085007-169085007 on Build GRCh38
rs926147748 Chr.3: 169085010-169085010 on Build GRCh38
rs760200964 Chr.3: 169085014-169085014 on Build GRCh38
rs780056793 Chr.3: 169085074-169085074 on Build GRCh38
rs772988182 Chr.3: 169084888-169084888 on Build GRCh38
rs766243412 Chr.3: 169084898-169084898 on Build GRCh38
rs946120209 Chr.3: 169085065-169085065 on Build GRCh38
rs769620374 Chr.3: 169084887-169084887 on Build GRCh38
rs757625757 Chr.3: 169084923-169084923 on Build GRCh38
rs200073118 Chr.3: 169084974-169084974 on Build GRCh38
rs765615508 Chr.3: 169085043-169085043 on Build GRCh38

View Full Product Details