Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67350086-67350337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMD12
Entrez Gene ID
5718
Gene Name
proteasome 26S subunit, non-ATPase 12
Gene Aliases
Rpn5, p55
UniGene
Hs.4295
Gene Chromosome Location
Chr.17: 67337916-67366627 on Build GRCh38
Transcripts
NM_001316341.1 NM_002816.4 NM_174871.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4187582 c.385C>T p.R129* Chr.17: 67350249-67350249 on Build GRCh38
4419688 c.368G>A p.R123Q Chr.17: 67350266-67350266 on Build GRCh38
707559 c.342A>T p.E114D Chr.17: 67350292-67350292 on Build GRCh38
4914724 c.399A>G p.E133E Chr.17: 67350235-67350235 on Build GRCh38
2797495 c.384A>G p.L128L Chr.17: 67350250-67350250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762351100 Chr.17: 67350195-67350195 on Build GRCh38
rs767936764 Chr.17: 67350266-67350266 on Build GRCh38
rs1025919595 Chr.17: 67350130-67350130 on Build GRCh38
rs34189633 Chr.17: 67350114-67350114 on Build GRCh38
rs1003481269 Chr.17: 67350186-67350186 on Build GRCh38
rs997044077 Chr.17: 67350224-67350224 on Build GRCh38
rs11870490 Chr.17: 67350211-67350211 on Build GRCh38
rs775859552 Chr.17: 67350276-67350276 on Build GRCh38
rs376420879 Chr.17: 67350274-67350274 on Build GRCh38
rs761043799 Chr.17: 67350297-67350297 on Build GRCh38
rs371518489 Chr.17: 67350181-67350181 on Build GRCh38
rs528046189 Chr.17: 67350137-67350137 on Build GRCh38
rs140191153 Chr.17: 67350301-67350301 on Build GRCh38
rs773069355 Chr.17: 67350214-67350216 on Build GRCh38
rs774683042 Chr.17: 67350232-67350232 on Build GRCh38
rs375216984 Chr.17: 67350189-67350189 on Build GRCh38
rs371724952 Chr.17: 67350238-67350238 on Build GRCh38
rs753998246 Chr.17: 67350306-67350306 on Build GRCh38
rs548572811 Chr.17: 67350163-67350163 on Build GRCh38
rs769405447 Chr.17: 67350182-67350183 on Build GRCh38
rs9897693 Chr.17: 67350278-67350278 on Build GRCh38
rs1021631123 Chr.17: 67350237-67350237 on Build GRCh38
rs769331852 Chr.17: 67350182-67350182 on Build GRCh38

View Full Product Details