Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30340432-30340701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM39
Entrez Gene ID
56658
Gene Name
tripartite motif containing 39
Gene Aliases
RNF23, TFP, TRIM39B
UniGene
Hs.413493
Gene Chromosome Location
Chr.6: 30326844-30343729 on Build GRCh38
Transcripts
NM_021253.3 NM_172016.2 NM_001199119.1

Gene Symbol
TRIM39-RPP21
Entrez Gene ID
202658
Gene Name
TRIM39-RPP21 readthrough
Gene Aliases
TRIM39R
UniGene
Hs.413493
Gene Chromosome Location
Chr.6: 30329311-30346858 on Build GRCh38
Transcripts
NM_021253.3 NM_172016.2 NM_001199119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5830395 c.866G>A p.S289N Chr.6: 30340567-30340567 on Build GRCh38
1443006 c.956G>A p.S319N Chr.6: 30340567-30340567 on Build GRCh38
6499333 c.954C>T p.F318F Chr.6: 30340565-30340565 on Build GRCh38
4160554 c.932C>G p.S311C Chr.6: 30340543-30340543 on Build GRCh38
4160556 c.842C>G p.S281C Chr.6: 30340543-30340543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767726574 Chr.6: 30340672-30340672 on Build GRCh38
rs763450349 Chr.6: 30340638-30340638 on Build GRCh38
rs746691124 Chr.6: 30340556-30340556 on Build GRCh38
rs370330384 Chr.6: 30340629-30340629 on Build GRCh38
rs762175442 Chr.6: 30340656-30340656 on Build GRCh38
rs929954900 Chr.6: 30340497-30340497 on Build GRCh38
rs752799369 Chr.6: 30340490-30340490 on Build GRCh38
rs1056866548 Chr.6: 30340567-30340567 on Build GRCh38
rs897379667 Chr.6: 30340511-30340511 on Build GRCh38
rs765751274 Chr.6: 30340591-30340591 on Build GRCh38
rs896510182 Chr.6: 30340568-30340568 on Build GRCh38
rs765694220 Chr.6: 30340489-30340489 on Build GRCh38
rs61758102 Chr.6: 30340549-30340549 on Build GRCh38
rs758418811 Chr.6: 30340500-30340500 on Build GRCh38
rs776890964 Chr.6: 30340574-30340574 on Build GRCh38
rs754890259 Chr.6: 30340539-30340539 on Build GRCh38
rs771773498 Chr.6: 30340546-30340546 on Build GRCh38
rs901209482 Chr.6: 30340488-30340488 on Build GRCh38
rs748534915 Chr.6: 30340478-30340478 on Build GRCh38
rs760001272 Chr.6: 30340486-30340486 on Build GRCh38
rs867800553 Chr.6: 30340667-30340667 on Build GRCh38
rs533132789 Chr.6: 30340468-30340468 on Build GRCh38
rs778936148 Chr.6: 30340541-30340541 on Build GRCh38
rs779752098 Chr.6: 30340520-30340520 on Build GRCh38
rs376126107 Chr.6: 30340639-30340639 on Build GRCh38
rs747666472 Chr.6: 30340543-30340543 on Build GRCh38
rs749067232 Chr.6: 30340530-30340530 on Build GRCh38
rs751798210 Chr.6: 30340514-30340514 on Build GRCh38
rs777033006 Chr.6: 30340472-30340472 on Build GRCh38
rs764307013 Chr.6: 30340505-30340505 on Build GRCh38
rs999989980 Chr.6: 30340661-30340661 on Build GRCh38
rs151317560 Chr.6: 30340570-30340570 on Build GRCh38
rs181089786 Chr.6: 30340664-30340664 on Build GRCh38
rs1003454172 Chr.6: 30340662-30340662 on Build GRCh38
rs759899884 Chr.6: 30340575-30340575 on Build GRCh38
rs529030096 Chr.6: 30340653-30340653 on Build GRCh38
rs757476654 Chr.6: 30340515-30340515 on Build GRCh38
rs772931363 Chr.6: 30340455-30340455 on Build GRCh38
rs753434203 Chr.6: 30340596-30340596 on Build GRCh38
rs760369976 Chr.6: 30340463-30340463 on Build GRCh38

View Full Product Details