Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35930453-35930724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RDM1
Entrez Gene ID
201299
Gene Name
RAD52 motif containing 1
Gene Aliases
RAD52B
UniGene
Hs.194411
Gene Chromosome Location
Chr.17: 35918080-35931821 on Build GRCh38
Transcripts
NM_001034836.1 NM_001163120.1 NM_001163121.1 NM_145654.3 NR_027996.1 NR_027997.1 NR_027998.1 XM_011524509.2 XM_011524510.2

SNP Detail

SNP ID Chromosome Location
rs774175904 Chr.17: 35930603-35930606 on Build GRCh38
rs746047907 Chr.17: 35930647-35930647 on Build GRCh38
rs751696852 Chr.17: 35930700-35930700 on Build GRCh38
rs375446393 Chr.17: 35930590-35930590 on Build GRCh38
rs770574507 Chr.17: 35930511-35930511 on Build GRCh38
rs561124314 Chr.17: 35930502-35930502 on Build GRCh38
rs781261386 Chr.17: 35930644-35930644 on Build GRCh38
rs771251798 Chr.17: 35930641-35930641 on Build GRCh38
rs757312370 Chr.17: 35930549-35930549 on Build GRCh38
rs909303950 Chr.17: 35930478-35930478 on Build GRCh38
rs753205627 Chr.17: 35930621-35930621 on Build GRCh38
rs544734058 Chr.17: 35930497-35930497 on Build GRCh38
rs554693641 Chr.17: 35930677-35930677 on Build GRCh38
rs753038483 Chr.17: 35930694-35930694 on Build GRCh38
rs778248130 Chr.17: 35930697-35930697 on Build GRCh38
rs377136088 Chr.17: 35930684-35930684 on Build GRCh38
rs768342943 Chr.17: 35930655-35930655 on Build GRCh38
rs143347588 Chr.17: 35930632-35930632 on Build GRCh38
rs546849615 Chr.17: 35930544-35930544 on Build GRCh38
rs554074570 Chr.17: 35930695-35930695 on Build GRCh38
rs191410149 Chr.17: 35930604-35930604 on Build GRCh38
rs11871113 Chr.17: 35930555-35930555 on Build GRCh38
rs747060291 Chr.17: 35930639-35930639 on Build GRCh38
rs372664878 Chr.17: 35930624-35930624 on Build GRCh38
rs2280786 Chr.17: 35930633-35930633 on Build GRCh38
rs902159014 Chr.17: 35930562-35930562 on Build GRCh38
rs765911906 Chr.17: 35930586-35930586 on Build GRCh38
rs370642067 Chr.17: 35930525-35930525 on Build GRCh38
rs141479106 Chr.17: 35930657-35930657 on Build GRCh38
rs943286995 Chr.17: 35930480-35930480 on Build GRCh38
rs201890775 Chr.17: 35930680-35930680 on Build GRCh38
rs779130250 Chr.17: 35930658-35930662 on Build GRCh38
rs760450291 Chr.17: 35930585-35930585 on Build GRCh38
rs188813804 Chr.17: 35930581-35930581 on Build GRCh38
rs772110798 Chr.17: 35930670-35930670 on Build GRCh38
rs761861258 Chr.17: 35930667-35930667 on Build GRCh38
rs73992632 Chr.17: 35930690-35930690 on Build GRCh38
rs760254079 Chr.17: 35930678-35930678 on Build GRCh38
rs942192509 Chr.17: 35930484-35930484 on Build GRCh38

View Full Product Details