Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21155651-21155886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5AU1
Entrez Gene ID
390445
Gene Name
olfactory receptor family 5 subfamily AU member 1
Gene Aliases
OR14-38
UniGene
Hs.553768
Gene Chromosome Location
Chr.14: 21154937-21156025 on Build GRCh38
Transcripts
NM_001004731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
469771 c.337C>T p.L113F Chr.14: 21155689-21155689 on Build GRCh38
954362 c.237C>T p.F79F Chr.14: 21155789-21155789 on Build GRCh38
4608513 c.274G>T p.G92W Chr.14: 21155752-21155752 on Build GRCh38
3814454 c.241G>A p.V81M Chr.14: 21155785-21155785 on Build GRCh38
1719815 c.234C>T p.L78L Chr.14: 21155792-21155792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140358816 Chr.14: 21155789-21155789 on Build GRCh38
rs764386262 Chr.14: 21155767-21155767 on Build GRCh38
rs45570932 Chr.14: 21155816-21155816 on Build GRCh38
rs775982725 Chr.14: 21155762-21155762 on Build GRCh38
rs765637237 Chr.14: 21155848-21155848 on Build GRCh38
rs749105137 Chr.14: 21155802-21155802 on Build GRCh38
rs773014178 Chr.14: 21155726-21155726 on Build GRCh38
rs771157723 Chr.14: 21155739-21155739 on Build GRCh38
rs111585524 Chr.14: 21155716-21155716 on Build GRCh38
rs551955955 Chr.14: 21155728-21155728 on Build GRCh38
rs751258580 Chr.14: 21155849-21155849 on Build GRCh38
rs79868143 Chr.14: 21155770-21155770 on Build GRCh38
rs767188314 Chr.14: 21155865-21155865 on Build GRCh38
rs775498204 Chr.14: 21155828-21155828 on Build GRCh38
rs17102042 Chr.14: 21155788-21155788 on Build GRCh38
rs952116920 Chr.14: 21155795-21155795 on Build GRCh38
rs776923357 Chr.14: 21155847-21155847 on Build GRCh38
rs759794584 Chr.14: 21155705-21155705 on Build GRCh38
rs202027012 Chr.14: 21155685-21155685 on Build GRCh38
rs932398181 Chr.14: 21155712-21155712 on Build GRCh38
rs767711572 Chr.14: 21155766-21155766 on Build GRCh38
rs767529086 Chr.14: 21155754-21155754 on Build GRCh38
rs150580198 Chr.14: 21155678-21155678 on Build GRCh38
rs4982419 Chr.14: 21155677-21155677 on Build GRCh38
rs535482449 Chr.14: 21155804-21155804 on Build GRCh38
rs761134951 Chr.14: 21155831-21155831 on Build GRCh38
rs746025698 Chr.14: 21155812-21155812 on Build GRCh38
rs976328746 Chr.14: 21155839-21155839 on Build GRCh38
rs17102038 Chr.14: 21155765-21155765 on Build GRCh38
rs202234533 Chr.14: 21155811-21155811 on Build GRCh38
rs943735169 Chr.14: 21155827-21155827 on Build GRCh38
rs150003696 Chr.14: 21155794-21155794 on Build GRCh38
rs368871939 Chr.14: 21155778-21155778 on Build GRCh38
rs991988563 Chr.14: 21155718-21155718 on Build GRCh38
rs200164964 Chr.14: 21155703-21155703 on Build GRCh38
rs376556611 Chr.14: 21155856-21155856 on Build GRCh38
rs139976871 Chr.14: 21155747-21155747 on Build GRCh38
rs1035831635 Chr.14: 21155683-21155683 on Build GRCh38
rs764630917 Chr.14: 21155842-21155842 on Build GRCh38
rs922264564 Chr.14: 21155690-21155690 on Build GRCh38
rs777472983 Chr.14: 21155798-21155798 on Build GRCh38
rs766122273 Chr.14: 21155773-21155773 on Build GRCh38

View Full Product Details