Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 64273366-64273631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCMTD2
Entrez Gene ID
55251
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2
Gene Aliases
C20orf36
UniGene
Hs.744845
Gene Chromosome Location
Chr.20: 64255695-64276226 on Build GRCh38
Transcripts
NM_001104925.1 NM_018257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6160674 c.274G>C p.E92Q Chr.20: 64273460-64273460 on Build GRCh38
5958723 c.1028delT p.L343fs*9 Chr.20: 64273542-64273542 on Build GRCh38
6741771 c.230G>A p.R77Q Chr.20: 64273416-64273416 on Build GRCh38
1615897 c.1080A>G p.E360E Chr.20: 64273594-64273594 on Build GRCh38
6160673 c.946G>C p.E316Q Chr.20: 64273460-64273460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866934581 Chr.20: 64273475-64273475 on Build GRCh38
rs149434467 Chr.20: 64273434-64273434 on Build GRCh38
rs750099571 Chr.20: 64273579-64273579 on Build GRCh38
rs368220103 Chr.20: 64273508-64273508 on Build GRCh38
rs867066957 Chr.20: 64273469-64273469 on Build GRCh38
rs767416315 Chr.20: 64273481-64273483 on Build GRCh38
rs928297627 Chr.20: 64273491-64273491 on Build GRCh38
rs200167050 Chr.20: 64273482-64273482 on Build GRCh38
rs762229962 Chr.20: 64273537-64273537 on Build GRCh38
rs146314236 Chr.20: 64273510-64273510 on Build GRCh38
rs375984565 Chr.20: 64273530-64273530 on Build GRCh38
rs914314593 Chr.20: 64273496-64273496 on Build GRCh38
rs559960598 Chr.20: 64273494-64273494 on Build GRCh38
rs542704294 Chr.20: 64273472-64273474 on Build GRCh38
rs200954397 Chr.20: 64273480-64273480 on Build GRCh38
rs765516486 Chr.20: 64273572-64273572 on Build GRCh38
rs916175141 Chr.20: 64273408-64273408 on Build GRCh38
rs372058955 Chr.20: 64273529-64273529 on Build GRCh38
rs776077457 Chr.20: 64273429-64273429 on Build GRCh38
rs757926855 Chr.20: 64273467-64273467 on Build GRCh38
rs144823949 Chr.20: 64273492-64273492 on Build GRCh38
rs557789541 Chr.20: 64273425-64273425 on Build GRCh38
rs994738031 Chr.20: 64273453-64273453 on Build GRCh38
rs747925562 Chr.20: 64273405-64273405 on Build GRCh38
rs777948252 Chr.20: 64273445-64273445 on Build GRCh38
rs779369325 Chr.20: 64273470-64273470 on Build GRCh38
rs772040163 Chr.20: 64273406-64273406 on Build GRCh38
rs750391743 Chr.20: 64273545-64273546 on Build GRCh38
rs759730113 Chr.20: 64273426-64273426 on Build GRCh38
rs868249434 Chr.20: 64273396-64273396 on Build GRCh38
rs542450173 Chr.20: 64273528-64273528 on Build GRCh38
rs781098622 Chr.20: 64273395-64273395 on Build GRCh38
rs751551512 Chr.20: 64273489-64273489 on Build GRCh38
rs771980963 Chr.20: 64273428-64273428 on Build GRCh38
rs529111736 Chr.20: 64273495-64273495 on Build GRCh38
rs764924217 Chr.20: 64273454-64273454 on Build GRCh38
rs762142179 Chr.20: 64273444-64273444 on Build GRCh38
rs761039003 Chr.20: 64273433-64273433 on Build GRCh38
rs772216700 Chr.20: 64273513-64273513 on Build GRCh38
rs914100599 Chr.20: 64273558-64273558 on Build GRCh38
rs750037182 Chr.20: 64273466-64273466 on Build GRCh38

View Full Product Details