Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 85996442-85996711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAA35
Entrez Gene ID
60560
Gene Name
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
Gene Aliases
EGAP, MAK10, MAK10P, bA379P1.1
UniGene
Hs.436098
Gene Chromosome Location
Chr.9: 85941142-86026355 on Build GRCh38
Transcripts
NM_001321881.1 NM_001321882.1 NM_024635.3 XM_005252127.3 XM_011518903.2 XM_011518904.2 XM_017015012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3927155 c.943C>T p.R315* Chr.9: 85996464-85996464 on Build GRCh38
1314996 c.993A>T p.R331S Chr.9: 85996514-85996514 on Build GRCh38
4375603 c.1022A>T p.E341V Chr.9: 85996543-85996543 on Build GRCh38
3220800 c.1045C>T p.H349Y Chr.9: 85996566-85996566 on Build GRCh38
4915949 c.974T>C p.M325T Chr.9: 85996495-85996495 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs928421958 Chr.9: 85996545-85996545 on Build GRCh38
rs753391670 Chr.9: 85996603-85996603 on Build GRCh38
rs938442464 Chr.9: 85996601-85996601 on Build GRCh38
rs774843999 Chr.9: 85996469-85996469 on Build GRCh38
rs746148077 Chr.9: 85996472-85996472 on Build GRCh38
rs910221060 Chr.9: 85996485-85996485 on Build GRCh38
rs752645607 Chr.9: 85996617-85996617 on Build GRCh38
rs199963761 Chr.9: 85996574-85996574 on Build GRCh38
rs527968784 Chr.9: 85996490-85996490 on Build GRCh38
rs1041946911 Chr.9: 85996669-85996669 on Build GRCh38
rs776668929 Chr.9: 85996534-85996534 on Build GRCh38
rs756971969 Chr.9: 85996604-85996604 on Build GRCh38
rs756092801 Chr.9: 85996584-85996584 on Build GRCh38
rs751374102 Chr.9: 85996548-85996548 on Build GRCh38
rs754673277 Chr.9: 85996550-85996550 on Build GRCh38
rs987061308 Chr.9: 85996478-85996478 on Build GRCh38
rs891951702 Chr.9: 85996617-85996617 on Build GRCh38
rs780435377 Chr.9: 85996626-85996626 on Build GRCh38
rs552213134 Chr.9: 85996612-85996612 on Build GRCh38
rs893416074 Chr.9: 85996587-85996587 on Build GRCh38
rs370054587 Chr.9: 85996611-85996611 on Build GRCh38
rs762082304 Chr.9: 85996541-85996541 on Build GRCh38
rs777477766 Chr.9: 85996590-85996590 on Build GRCh38
rs770391227 Chr.9: 85996646-85996646 on Build GRCh38
rs568890931 Chr.9: 85996645-85996645 on Build GRCh38
rs772643338 Chr.9: 85996476-85996476 on Build GRCh38
rs200918536 Chr.9: 85996522-85996522 on Build GRCh38
rs760774532 Chr.9: 85996497-85996497 on Build GRCh38
rs532139527 Chr.9: 85996575-85996575 on Build GRCh38
rs765375299 Chr.9: 85996546-85996546 on Build GRCh38
rs1012257102 Chr.9: 85996670-85996670 on Build GRCh38
rs772283551 Chr.9: 85996625-85996625 on Build GRCh38
rs973931955 Chr.9: 85996465-85996465 on Build GRCh38

View Full Product Details