Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60282800-60283041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPM1A
Entrez Gene ID
5494
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases
PP2C-ALPHA, PP2CA, PP2Calpha
UniGene
Hs.130036
Gene Chromosome Location
Chr.14: 60245752-60299087 on Build GRCh38
Transcripts
NM_021003.4 NM_177951.2 NM_177952.2 XM_005267779.1 XM_005267781.1 XM_011536872.1 XM_011536876.1 XM_011536877.1 XM_011536878.1 XM_011536879.2 XM_011536880.2 XM_011536881.2 XM_011536882.2 XM_011536883.1 XM_011536884.1 XM_017021381.1 XM_017021382.1 XM_017021383.1 XM_017021384.1 XM_017021385.1 XM_017021386.1 XM_017021387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6050437 c.123G>A p.T41T Chr.14: 60282826-60282826 on Build GRCh38
6337879 c.534T>C p.F178F Chr.14: 60283018-60283018 on Build GRCh38
3815019 c.194C>G p.S65C Chr.14: 60282897-60282897 on Build GRCh38
1300717 c.293A>G p.K98R Chr.14: 60282996-60282996 on Build GRCh38
6140791 c.555G>A p.M185I Chr.14: 60283039-60283039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745312893 Chr.14: 60282944-60282944 on Build GRCh38
rs1006147517 Chr.14: 60282909-60282909 on Build GRCh38
rs755965320 Chr.14: 60282918-60282918 on Build GRCh38
rs766728463 Chr.14: 60282994-60282994 on Build GRCh38
rs773920259 Chr.14: 60282940-60282940 on Build GRCh38
rs149289409 Chr.14: 60282859-60282859 on Build GRCh38
rs576909532 Chr.14: 60282892-60282892 on Build GRCh38
rs765800676 Chr.14: 60282834-60282834 on Build GRCh38
rs773398411 Chr.14: 60282979-60282979 on Build GRCh38
rs771770007 Chr.14: 60282960-60282960 on Build GRCh38
rs758458583 Chr.14: 60282836-60282836 on Build GRCh38
rs755806358 Chr.14: 60282858-60282858 on Build GRCh38
rs559528140 Chr.14: 60282935-60282935 on Build GRCh38
rs766500262 Chr.14: 60282845-60282845 on Build GRCh38
rs775119723 Chr.14: 60282964-60282964 on Build GRCh38
rs750913302 Chr.14: 60282835-60282835 on Build GRCh38
rs199939814 Chr.14: 60282939-60282939 on Build GRCh38
rs777894788 Chr.14: 60282926-60282926 on Build GRCh38
rs375823365 Chr.14: 60282846-60282846 on Build GRCh38
rs767093859 Chr.14: 60282826-60282826 on Build GRCh38
rs376150210 Chr.14: 60282835-60282836 on Build GRCh38
rs952088786 Chr.14: 60282980-60282980 on Build GRCh38
rs117800726 Chr.14: 60282931-60282931 on Build GRCh38
rs759267033 Chr.14: 60282821-60282821 on Build GRCh38
rs995311863 Chr.14: 60282883-60282883 on Build GRCh38
rs199565515 Chr.14: 60282833-60282833 on Build GRCh38
rs763545516 Chr.14: 60282981-60282981 on Build GRCh38
rs145211997 Chr.14: 60282978-60282978 on Build GRCh38
rs74713917 Chr.14: 60282831-60282831 on Build GRCh38

View Full Product Details