Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43670670-43670931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPH9
Entrez Gene ID
221421
Gene Name
radial spoke head 9 homolog
Gene Aliases
C6orf206, CILD12, MRPS18AL1
UniGene
Hs.534585
Gene Chromosome Location
Chr.6: 43645030-43671011 on Build GRCh38
Transcripts
NM_001193341.1 NM_152732.4 XM_005248901.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3874388 c.718C>T p.R240C Chr.6: 43670836-43670836 on Build GRCh38
3874387 c.770C>T p.A257V Chr.6: 43670836-43670836 on Build GRCh38
4962992 c.835C>T p.L279L Chr.6: 43670901-43670901 on Build GRCh38
6174541 c.696G>A p.R232R Chr.6: 43670814-43670814 on Build GRCh38
1079669 c.777_779delCTA p.Y260delY Chr.6: 43670895-43670897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749404508 Chr.6: 43670838-43670838 on Build GRCh38
rs368354393 Chr.6: 43670768-43670768 on Build GRCh38
rs777862194 Chr.6: 43670909-43670909 on Build GRCh38
rs953128048 Chr.6: 43670708-43670708 on Build GRCh38
rs996840893 Chr.6: 43670788-43670788 on Build GRCh38
rs780394947 Chr.6: 43670891-43670891 on Build GRCh38
rs1057502519 Chr.6: 43670822-43670822 on Build GRCh38
rs769469438 Chr.6: 43670853-43670853 on Build GRCh38
rs16896629 Chr.6: 43670899-43670899 on Build GRCh38
rs372218633 Chr.6: 43670898-43670898 on Build GRCh38
rs894482674 Chr.6: 43670852-43670852 on Build GRCh38
rs781527232 Chr.6: 43670739-43670739 on Build GRCh38
rs748165900 Chr.6: 43670845-43670845 on Build GRCh38
rs550766275 Chr.6: 43670744-43670744 on Build GRCh38
rs566804409 Chr.6: 43670815-43670815 on Build GRCh38
rs774350226 Chr.6: 43670758-43670758 on Build GRCh38
rs376190455 Chr.6: 43670818-43670818 on Build GRCh38
rs753489926 Chr.6: 43670801-43670801 on Build GRCh38
rs200193470 Chr.6: 43670878-43670878 on Build GRCh38
rs756476977 Chr.6: 43670880-43670880 on Build GRCh38
rs763265177 Chr.6: 43670741-43670745 on Build GRCh38
rs774281454 Chr.6: 43670843-43670843 on Build GRCh38
rs200144701 Chr.6: 43670867-43670867 on Build GRCh38
rs558538408 Chr.6: 43670863-43670863 on Build GRCh38
rs760442254 Chr.6: 43670784-43670784 on Build GRCh38
rs757687268 Chr.6: 43670820-43670820 on Build GRCh38
rs371417111 Chr.6: 43670861-43670861 on Build GRCh38
rs746265610 Chr.6: 43670823-43670823 on Build GRCh38
rs754145780 Chr.6: 43670892-43670892 on Build GRCh38
rs772227123 Chr.6: 43670905-43670905 on Build GRCh38
rs548178595 Chr.6: 43670785-43670785 on Build GRCh38
rs570578504 Chr.6: 43670864-43670864 on Build GRCh38
rs768848964 Chr.6: 43670841-43670841 on Build GRCh38
rs754525382 Chr.6: 43670813-43670813 on Build GRCh38
rs749410104 Chr.6: 43670749-43670749 on Build GRCh38
rs756423549 Chr.6: 43670836-43670836 on Build GRCh38
rs761650356 Chr.6: 43670760-43670760 on Build GRCh38
rs374243816 Chr.6: 43670750-43670750 on Build GRCh38
rs764582054 Chr.6: 43670814-43670814 on Build GRCh38
rs78970418 Chr.6: 43670748-43670748 on Build GRCh38
rs768034790 Chr.6: 43670879-43670879 on Build GRCh38
rs758225342 Chr.6: 43670832-43670832 on Build GRCh38
rs746140683 Chr.6: 43670740-43670740 on Build GRCh38
rs375971288 Chr.6: 43670876-43670876 on Build GRCh38
rs371183985 Chr.6: 43670781-43670781 on Build GRCh38
rs375121621 Chr.6: 43670904-43670904 on Build GRCh38
rs143815397 Chr.6: 43670837-43670837 on Build GRCh38
rs900714215 Chr.6: 43670786-43670786 on Build GRCh38
rs773242367 Chr.6: 43670851-43670851 on Build GRCh38
rs772102571 Chr.6: 43670847-43670847 on Build GRCh38

View Full Product Details