Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31630634-31630879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCRTR1
Entrez Gene ID
3061
Gene Name
hypocretin receptor 1
Gene Aliases
OX1R
UniGene
Hs.388226
Gene Chromosome Location
Chr.1: 31617700-31632518 on Build GRCh38
Transcripts
XM_017001107.1 NM_012392.3 NR_033686.1 NR_033688.1 XM_011541745.1 XM_011541746.2 XM_011541747.1 XM_017001680.1 XM_017001681.1

Gene Symbol
PEF1
Entrez Gene ID
553115
Gene Name
penta-EF-hand domain containing 1
Gene Aliases
ABP32, PEF1A
UniGene
Hs.470417
Gene Chromosome Location
Chr.1: 31629862-31645237 on Build GRCh38
Transcripts
XM_017001107.1 NM_012392.3 NR_033686.1 NR_033688.1 XM_011541745.1 XM_011541746.2 XM_011541747.1 XM_017001680.1 XM_017001681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6881711 c.787G>A p.A263T Chr.1: 31630681-31630681 on Build GRCh38
2077211 c.787G>T p.A263S Chr.1: 31630681-31630681 on Build GRCh38
7194220 c.817G>A p.E273K Chr.1: 31630651-31630651 on Build GRCh38
5809598 c.689G>A p.C230Y Chr.1: 31630779-31630779 on Build GRCh38
3746933 c.804T>C p.I268I Chr.1: 31630664-31630664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555613785 Chr.1: 31630796-31630796 on Build GRCh38
rs749213335 Chr.1: 31630678-31630678 on Build GRCh38
rs866571271 Chr.1: 31630768-31630768 on Build GRCh38
rs778826617 Chr.1: 31630820-31630820 on Build GRCh38
rs1029317443 Chr.1: 31630808-31630808 on Build GRCh38
rs774191997 Chr.1: 31630690-31630690 on Build GRCh38
rs201297417 Chr.1: 31630836-31630836 on Build GRCh38
rs374586693 Chr.1: 31630833-31630833 on Build GRCh38
rs187435521 Chr.1: 31630835-31630835 on Build GRCh38
rs766788613 Chr.1: 31630769-31630769 on Build GRCh38
rs765758652 Chr.1: 31630750-31630750 on Build GRCh38
rs1016119085 Chr.1: 31630856-31630856 on Build GRCh38
rs779812577 Chr.1: 31630711-31630712 on Build GRCh38
rs1061770 Chr.1: 31630664-31630664 on Build GRCh38
rs549808725 Chr.1: 31630756-31630756 on Build GRCh38
rs780909453 Chr.1: 31630747-31630747 on Build GRCh38
rs755186565 Chr.1: 31630774-31630774 on Build GRCh38
rs775229794 Chr.1: 31630851-31630851 on Build GRCh38
rs780416054 Chr.1: 31630658-31630658 on Build GRCh38
rs953350614 Chr.1: 31630816-31630816 on Build GRCh38
rs745659069 Chr.1: 31630693-31630693 on Build GRCh38
rs569702331 Chr.1: 31630759-31630759 on Build GRCh38
rs776178785 Chr.1: 31630744-31630744 on Build GRCh38
rs781284157 Chr.1: 31630675-31630675 on Build GRCh38
rs768631094 Chr.1: 31630668-31630668 on Build GRCh38
rs1019932391 Chr.1: 31630739-31630739 on Build GRCh38
rs547542586 Chr.1: 31630779-31630779 on Build GRCh38
rs760275627 Chr.1: 31630707-31630707 on Build GRCh38
rs371471126 Chr.1: 31630857-31630857 on Build GRCh38
rs775137790 Chr.1: 31630696-31630696 on Build GRCh38
rs757209663 Chr.1: 31630805-31630805 on Build GRCh38
rs985013441 Chr.1: 31630832-31630832 on Build GRCh38
rs539499336 Chr.1: 31630681-31630681 on Build GRCh38
rs763547860 Chr.1: 31630743-31630743 on Build GRCh38
rs371830019 Chr.1: 31630773-31630773 on Build GRCh38
rs139896724 Chr.1: 31630785-31630785 on Build GRCh38
rs771880451 Chr.1: 31630845-31630845 on Build GRCh38
rs758834965 Chr.1: 31630764-31630764 on Build GRCh38
rs901516625 Chr.1: 31630772-31630772 on Build GRCh38
rs146006007 Chr.1: 31630695-31630695 on Build GRCh38

View Full Product Details