Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76473520-76473753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBDF2
Entrez Gene ID
79651
Gene Name
rhomboid 5 homolog 2
Gene Aliases
RHBDL5, RHBDL6, TEC, TOC, TOCG, iRhom2
UniGene
Hs.464157
Gene Chromosome Location
Chr.17: 76470893-76501440 on Build GRCh38
Transcripts
NM_001005498.3 NM_024599.5 XM_005257669.3 XM_005257670.1 XM_011525249.2 XM_011525250.2 XM_011525251.2 XM_017025079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398528 c.1753C>A p.H585N Chr.17: 76473715-76473715 on Build GRCh38
6399914 c.1799G>T p.C600F Chr.17: 76473669-76473669 on Build GRCh38
4271568 c.1780G>A p.E594K Chr.17: 76473688-76473688 on Build GRCh38
1195190 c.1730G>A p.C577Y Chr.17: 76473738-76473738 on Build GRCh38
6224637 c.1779C>T p.C593C Chr.17: 76473689-76473689 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371381813 Chr.17: 76473603-76473603 on Build GRCh38
rs182709534 Chr.17: 76473675-76473675 on Build GRCh38
rs189809413 Chr.17: 76473641-76473641 on Build GRCh38
rs371731107 Chr.17: 76473656-76473656 on Build GRCh38
rs747526150 Chr.17: 76473645-76473645 on Build GRCh38
rs374276167 Chr.17: 76473608-76473608 on Build GRCh38
rs770029904 Chr.17: 76473605-76473605 on Build GRCh38
rs767104777 Chr.17: 76473612-76473612 on Build GRCh38
rs942171489 Chr.17: 76473581-76473581 on Build GRCh38
rs551896954 Chr.17: 76473542-76473542 on Build GRCh38
rs923251817 Chr.17: 76473551-76473551 on Build GRCh38
rs552399716 Chr.17: 76473603-76473604 on Build GRCh38
rs370621586 Chr.17: 76473614-76473614 on Build GRCh38
rs760264421 Chr.17: 76473721-76473721 on Build GRCh38
rs111548723 Chr.17: 76473597-76473597 on Build GRCh38
rs750748859 Chr.17: 76473630-76473630 on Build GRCh38
rs772158066 Chr.17: 76473699-76473699 on Build GRCh38
rs143001224 Chr.17: 76473724-76473724 on Build GRCh38
rs73998915 Chr.17: 76473697-76473697 on Build GRCh38
rs773420212 Chr.17: 76473688-76473688 on Build GRCh38
rs976134127 Chr.17: 76473543-76473543 on Build GRCh38
rs762906914 Chr.17: 76473701-76473701 on Build GRCh38
rs1052989151 Chr.17: 76473628-76473628 on Build GRCh38
rs983668258 Chr.17: 76473564-76473564 on Build GRCh38
rs367778886 Chr.17: 76473642-76473642 on Build GRCh38
rs368446500 Chr.17: 76473610-76473610 on Build GRCh38
rs909390356 Chr.17: 76473566-76473566 on Build GRCh38
rs781205088 Chr.17: 76473661-76473661 on Build GRCh38
rs948460041 Chr.17: 76473604-76473604 on Build GRCh38
rs770738781 Chr.17: 76473692-76473692 on Build GRCh38
rs571664645 Chr.17: 76473555-76473555 on Build GRCh38
rs535334733 Chr.17: 76473689-76473689 on Build GRCh38
rs772937305 Chr.17: 76473606-76473606 on Build GRCh38
rs533947189 Chr.17: 76473627-76473627 on Build GRCh38
rs763851676 Chr.17: 76473622-76473622 on Build GRCh38
rs775082312 Chr.17: 76473711-76473711 on Build GRCh38
rs755778618 Chr.17: 76473619-76473619 on Build GRCh38
rs34946217 Chr.17: 76473686-76473687 on Build GRCh38
rs748243976 Chr.17: 76473662-76473662 on Build GRCh38
rs759443048 Chr.17: 76473611-76473611 on Build GRCh38

View Full Product Details