Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241107948-241108205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PASK
Entrez Gene ID
23178
Gene Name
PAS domain containing serine/threonine kinase
Gene Aliases
PASKIN, STK37
UniGene
Hs.397891
Gene Chromosome Location
Chr.2: 241106099-241150347 on Build GRCh38
Transcripts
NM_001252119.1 NM_001252120.1 NM_001252122.1 NM_015148.3 XM_005246991.1 XM_011510826.1 XM_011510828.1 XM_011510829.1 XM_011510830.1 XM_011510831.2 XM_011510832.2 XM_011510833.1 XM_011510834.1 XM_011510835.1 XM_017003634.1 XM_017003635.1 XM_017003636.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1018817 c.3648G>A p.P1216P Chr.2: 241108186-241108186 on Build GRCh38
4777423 c.3629T>G p.V1210G Chr.2: 241108205-241108205 on Build GRCh38
4133705 c.3644A>C p.H1215P Chr.2: 241108190-241108190 on Build GRCh38
6738069 c.3647C>T p.P1216L Chr.2: 241108187-241108187 on Build GRCh38
2910129 c.3649C>T p.P1217S Chr.2: 241108185-241108185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34777851 Chr.2: 241108115-241108115 on Build GRCh38
rs1004636726 Chr.2: 241108022-241108022 on Build GRCh38
rs769833623 Chr.2: 241108187-241108187 on Build GRCh38
rs367678856 Chr.2: 241108137-241108137 on Build GRCh38
rs887710146 Chr.2: 241108020-241108020 on Build GRCh38
rs748534676 Chr.2: 241108145-241108145 on Build GRCh38
rs983167318 Chr.2: 241108023-241108023 on Build GRCh38
rs781027284 Chr.2: 241108136-241108136 on Build GRCh38
rs1038814121 Chr.2: 241108072-241108072 on Build GRCh38
rs563432464 Chr.2: 241108166-241108166 on Build GRCh38
rs751415889 Chr.2: 241108128-241108128 on Build GRCh38
rs568076515 Chr.2: 241108009-241108009 on Build GRCh38
rs749730466 Chr.2: 241108149-241108150 on Build GRCh38
rs891742948 Chr.2: 241108111-241108111 on Build GRCh38
rs533854544 Chr.2: 241108123-241108123 on Build GRCh38
rs992824604 Chr.2: 241107978-241107978 on Build GRCh38
rs900351327 Chr.2: 241108103-241108104 on Build GRCh38
rs776888105 Chr.2: 241108179-241108179 on Build GRCh38
rs756924771 Chr.2: 241108130-241108130 on Build GRCh38
rs772645211 Chr.2: 241108156-241108156 on Build GRCh38
rs371770560 Chr.2: 241108160-241108160 on Build GRCh38
rs186279111 Chr.2: 241108055-241108055 on Build GRCh38
rs59710610 Chr.2: 241108104-241108104 on Build GRCh38
rs200123709 Chr.2: 241108186-241108186 on Build GRCh38
rs762804923 Chr.2: 241108121-241108121 on Build GRCh38
rs977584043 Chr.2: 241108150-241108150 on Build GRCh38
rs755633365 Chr.2: 241108139-241108139 on Build GRCh38
rs573511553 Chr.2: 241108141-241108142 on Build GRCh38
rs535513035 Chr.2: 241108043-241108043 on Build GRCh38
rs771109982 Chr.2: 241108161-241108161 on Build GRCh38
rs771161412 Chr.2: 241108002-241108002 on Build GRCh38
rs1034341044 Chr.2: 241108113-241108113 on Build GRCh38
rs550675198 Chr.2: 241108048-241108048 on Build GRCh38
rs918216363 Chr.2: 241107982-241107982 on Build GRCh38
rs752634792 Chr.2: 241108120-241108120 on Build GRCh38
rs574515379 Chr.2: 241108144-241108144 on Build GRCh38
rs1033810896 Chr.2: 241108174-241108174 on Build GRCh38

View Full Product Details